Skip to Content
Merck
All Photos(1)

Documents

PHR1124

Supelco

Myristic acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1-Tridecanecarboxylic acid, C14:0, NSC 5028, Tetradecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOH
CAS Number:
Molecular Weight:
228.37
Beilstein:
508624
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1448990

API family

myristic acid

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

250 °C/100 mmHg (lit.)

mp

52-54 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCCCCCCCCCCCCC(O)=O

InChI

1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)

InChI key

TUNFSRHWOTWDNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Myristic acid is a saturated fatty acid, which can increase the low-density lipoprotein cholesterol level and its sources are found to be coconut oil, palm kernal oil, butter fat and nutmeg.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7148 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Health Effects of Dietary Fatty Acids (1991)
Principles of Medical Biochemistry E-Book (2011)
Dalit Shental-Bechor et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(44), 17839-17844 (2012-08-01)
We present an integrated experimental and computational study of the molecular mechanisms by which myristoylation affects protein folding and function, which has been little characterized to date. Myristoylation, the covalent linkage of a hydrophobic C14 fatty acyl chain to the
Anja Meier et al.
Hepatology (Baltimore, Md.), 58(1), 31-42 (2012-12-06)
Chronic infection with the human hepatitis B virus (HBV) is a global health problem and a main cause of progressive liver diseases. HBV exhibits a narrow host range, replicating primarily in hepatocytes. Both host and hepatocyte specificity presumably involve specific
S Curry et al.
Biochimica et biophysica acta, 1441(2-3), 131-140 (1999-11-26)
Human serum albumin possesses multiple fatty acid binding sites of varying affinities, but the precise locations of these sites have remained elusive. The determination of the crystal structure of human serum albumin complexed with myristic acid recently revealed the positions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service