Skip to Content
Merck
All Photos(1)

Key Documents

PHR1118

Supelco

Propyl gallate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

PG, 3,4,5-Trihydroxybenzoic acid propyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4,5-(HO)3C6H2CO2CH2CH2CH3
CAS Number:
Molecular Weight:
212.20
Beilstein:
1877976
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E310
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. P3640000
traceable to USP 1576800

API family

propyl gallate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

146-149 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCCOC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3

InChI key

ZTHYODDOHIVTJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Propyl gallate is a synthetic phenolic antioxidant (SPA) utilized in food additives to inhibit or regulate the lipid oxidation. SPAs are cost-effective, largely available and highly efficient in comparison to their natural counterparts.

Application

Propyl gallate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and fatty liquids by chromatography and spectrophotometry.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Reconstitution

Prepare a 0.1M solution in glycerol:PBS (9:1)

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8981 in the slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

368.6 °F - closed cup

Flash Point(C)

187 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method validation and measurement uncertainty for the simultaneous determination of synthetic phenolic antioxidants in edible oils commonly consumed in Korea
Kim JM, et al.
Food Chemistry, 213(6), 19-25 (2016)
Determination of some antioxidants by derivative spectrophotometric method and high-performance liquid chromatography
Ivanovic D, et al.
Analytical Letters, 23(6), 1123-1137 (1990)
Detection and quantification of low-molecular-weight aldehydes in pharmaceutical excipients by headspace gas chromatography
Li Z, et al.
Journal of Chromatography A, 1104(1-2), 1-10 (2006)
Paola Colindres et al.
Journal of the science of food and agriculture, 91(5), 963-968 (2011-01-22)
The effect of selected antioxidants (grape seed extract (GS), oleoresin rosemary (OR), water-soluble oregano extract (WO), propyl gallate (PG), butylated hydroxyanisole (BHA)), butylated hydroxytoluene (BHT)) on sensory, color and oxidative stability of cooked, frozen, reheated ground beef patties was evaluated.
Brianne A Wilton et al.
Virology, 374(1), 82-99 (2008-01-29)
Cellular proteins containing BTB and kelch domains have been shown to function as adapters for the recruitment of substrates to cullin-3-based ubiquitin ligases. Poxviruses are the only family of viruses known to encode multiple BTB/kelch proteins, suggesting that poxviruses may

Articles

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

HPLC-UV method detects 9 antioxidants like propyl gallate in food using a Discovery®

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service