Skip to Content
Merck
All Photos(3)

Key Documents

P3130

Sigma-Aldrich

Propyl gallate

≥98% (HPLC), powder, antioxidant

Synonym(s):

3,4,5-Trihydroxybenzoic acid propyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4,5-(HO)3C6H2CO2CH2CH2CH3
CAS Number:
Molecular Weight:
212.20
Beilstein:
1877976
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
39024506
PubChem Substance ID:
NACRES:
NA.77

Product Name

Propyl gallate, powder

Assay

≥98%

Quality Level

form

powder

mp

146-149 °C (lit.)

solubility

ethanol: 50 mg/mL

SMILES string

CCCOC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3

InChI key

ZTHYODDOHIVTJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Reconstitution

Prepare a 0.1M solution in glycerol:PBS (9:1)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

368.6 °F - closed cup

Flash Point(C)

187 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Schuld et al.
Acta neuropathologica communications, 8(1), 154-154 (2020-09-06)
Filamin C (FLNc) is mainly expressed in striated muscle cells where it localizes to Z-discs, myotendinous junctions and intercalated discs. Recent studies have revealed numerous mutations in the FLNC gene causing familial and sporadic myopathies and cardiomyopathies with marked clinical
Lynn M Boyden et al.
The Journal of investigative dermatology, 135(6), 1540-1547 (2014-11-15)
Genetic investigation of inherited skin disorders has informed the understanding of skin self-renewal, differentiation, and barrier function. Erythrokeratodermia variabilis et progressiva (EKVP) is a rare, inherited skin disease that is characterized by transient figurate patches of erythema, localized or generalized
Carlas S Smith et al.
Nature communications, 10(1), 272-272 (2019-01-19)
Single-molecule binding assays enable the study of how molecular machines assemble and function. Current algorithms can identify and locate individual molecules, but require tedious manual validation of each spot. Moreover, no solution for high-throughput analysis of single-molecule binding data exists.
Mette Flinck et al.
Cells, 9(2) (2020-02-23)
Increased metabolic acid production and upregulation of net acid extrusion render pH homeostasis profoundly dysregulated in many cancers. Plasma membrane activity of vacuolar H+ ATPases (V-ATPases) has been implicated in acid extrusion and invasiveness of some cancers, yet often on
Matloob Ahmad et al.
European journal of medicinal chemistry, 45(2), 698-704 (2009-12-08)
A series of potential anti-oxidant and anti-bacterial N'-arylmethylidene-2-(3,4-dimethyl-5,5-dioxidopyrazolo[4,3-c][1,2]benzothiazin-2(4H)-yl)acetohydrazides was synthesized in a facile way starting from commercially available saccharine. 3,4-Dimethyl-2,4-dihydropyrazolo[4,3-c][1,2]benzothiazine 5,5-dioxide was obtained by ring expansion of 2-(2-oxopropyl)-1,2-benzisothiazol-3(2H)-one 1,1-dioxide followed by N-methylation and cyclization with hydrazine using ultrasonic irradiation. N-alkylation

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service