Skip to Content
Merck
All Photos(1)

Documents

76170

Supelco

Melting point standard 121-123°C

analytical standard

Synonym(s):

Benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12
Beilstein:
636131
EC Number:
MDL number:
UNSPSC Code:
20121904
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.21 (vs air)

vapor pressure

10 mmHg ( 132 °C)

autoignition temp.

1061 °F

shelf life

limited shelf life, expiry date on the label

bp

249 °C (lit.)

mp

121-123 °C (±0.3°C)
121-125 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

OC(=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoic acid melting point standard, 121-123 °C is useful as a melting point calibration standard while defining the purity of pharmaceutical chemicals. Its melting point value is determined with an average of 6 to 12 measurements using a Büchi B-545 instrument that is calibrated against primary standards.

Application

Melting point standard 121-123 °C, benzoic acid is an analytical standard used to calibrate melting point instruments, in the thermodynamic mode of operation. It is also used in R & D labs for chemical quality control (QC) and quality assurance (QA) to identify solid crystalline substances and to check their purity.

Features and Benefits

  • Melting point calibration standard traceable to primary standards (LGC, London)
  • Grade: Analytical Standard
  • Uncertainty of measurement up to ± 0.3 °C
  • Provided with certificates of analysis and safety data sheet
  • A product of analytical standard grade that meets the QC/QA requirements of melting point analysis

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kasture.V.A
Pharmaceutical Analysis, 1 (2008)
Ly Dieu Tran et al.
Journal of the American Chemical Society, 134(44), 18237-18240 (2012-10-30)
An auxiliary-assisted, copper catalyzed or promoted sulfenylation of benzoic acid derivative β-C-H bonds and benzylamine derivative γ-C-H bonds has been developed. The method employs disulfide reagents, copper(II) acetate, and DMSO solvent at 90-130 °C. Application of this methodology to the
Copper-mediated C-H/C-H biaryl coupling of benzoic acid derivatives and 1,3-azoles.
Mayuko Nishino et al.
Angewandte Chemie (International ed. in English), 52(16), 4457-4461 (2013-03-21)
Wanchun Xiang et al.
ChemSusChem, 6(2), 256-260 (2013-01-25)
Sensing the sun: Incorporation of a cyanomethyl benzoic acid electron acceptor into donor-π-acceptor sensitizers for dye-sensitized-solar cell is shown to lead to devices with improved conversion efficiency when compared with more widely used cyanoacetic acid acceptor.
Renbo Wei et al.
Macromolecular rapid communications, 34(4), 330-334 (2013-01-03)
A new approach is developed to fabricate highly oriented mono-domain LCE nano/microstructures through micro-molding in capillaries. Gratings and microwires as two typical examples are fabricated and characterized by polarizing optical microscopy, optical microscopy, and scanning electron microscopy. The gratings with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service