Skip to Content
Merck
All Photos(1)

Key Documents

P1800000

Pindolol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-(1H-Indol-4-yloxy)-3-(isopropylamino)-2-propanol, 1-(1H-Indol-4-yloxy)-3-[(1-methylethyl)amino]-2-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O2
CAS Number:
Molecular Weight:
248.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pindolol

manufacturer/tradename

EDQM

mp

167-171 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)NCC(O)COc1cccc2[nH]ccc12

InChI

1S/C14H20N2O2/c1-10(2)16-8-11(17)9-18-14-5-3-4-13-12(14)6-7-15-13/h3-7,10-11,15-17H,8-9H2,1-2H3

InChI key

JZQKKSLKJUAGIC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pindolol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

β1-adrenoceptor antagonist; putative 5-HT1A serotonin receptor agonist; vasodilator.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S Olver et al.
The Australian and New Zealand journal of psychiatry, 34(1), 71-79 (2001-02-24)
To critically review the literature on clinical trials in which pindolol, a 5HT1A receptor antagonist, has been used to augment the effects of antidepressants in patients with depression and to examine the pharmacodynamics and pharmacokinetics that may underlie such augmentations.
Drug therapy. Pindolol: a new beta-adrenoceptor antagonist with partial agonist activity.
W H Frishman
The New England journal of medicine, 308(16), 940-944 (1983-04-21)
Antihypertensive effects of pindolol.
L M Gonasun
American heart journal, 104(2 Pt 2), 374-387 (1982-08-01)
P Blier et al.
The Journal of clinical psychiatry, 59 Suppl 5, 16-23 (1998-06-27)
Serotonin (5-HT) selective reuptake inhibitors (SSRIs) and monoamine oxidase inhibitors are thought to have a delayed onset of antidepressant action attributable in part to the decrease in firing activity of 5-HT neurons they produce upon treatment initiation. As cell body
W H Aellig
American heart journal, 104(2 Pt 2), 346-356 (1982-08-01)
Beta-adrenoceptor blockade is responsible for the therapeutic action of beta-adrenoceptor-blocking drugs in the treatment of hypertension and angina pectoris. Many aspects of their effects can therefore be studied in relatively simple clinical pharmacologic experiments. Cardiac beta-adrenoceptor blockade can be measured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service