Skip to Content
Merck
All Photos(1)

Key Documents

OG100

Supelco

Oil and Grease 100 mg/L Calibration Standard

certified reference material, in H2O

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

CofA

certificate is enclosed in each package

manufacturer/tradename

RTC OG100-250

concentration

in H2O

application(s)

environmental
food and beverages

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service