Skip to Content
Merck
All Photos(1)

Key Documents

O5879

Supelco

Ornidazole

analytical standard

Synonym(s):

1-(3-Chloro-2-hydroxypropyl)-2-methyl-5-nitroimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H10N3O3Cl
CAS Number:
Molecular Weight:
219.63
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cc1ncc(n1CC(O)CCl)[N+]([O-])=O

InChI

1S/C7H10ClN3O3/c1-5-9-3-7(11(13)14)10(5)4-6(12)2-8/h3,6,12H,2,4H2,1H3

InChI key

IPWKIXLWTCNBKN-UHFFFAOYSA-N

Application

Ornidazole has been used as a standard in the determination of traces of ornidazole in samples of aquaculture tissue using ultra high performance liquid chromatography coupled with mass spectrometry (UHPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subhash S Vaghani et al.
Carbohydrate research, 347(1), 76-82 (2011-11-22)
In the present study, carboxymethyl chitosan was prepared from chitosan, crosslinked with glutaraldehyde and evaluated in vitro as a potential carrier for colon targeted drug delivery of ornidazole. Ornidazole was incorporated at the time of crosslinking of carboxymethyl chitosan. The
Kriti Shivhare et al.
International journal of biological macromolecules, 106, 775-783 (2017-08-19)
Molecular self-assembly of biodegradable amphiphilic polymers allows rational design of biocompatible nanomaterials for drug delivery. Use of substituted polysaccharides for such applications offers the ease of design and synthesis, and provides higher biofunctionality and biocompatibility to nanomaterials. The present work
Nathalie Mahé et al.
Journal of pharmaceutical sciences, 100(6), 2258-2266 (2011-01-20)
The solid-state properties of antiprotozoal ternidazole (3-(2-methyl-5-nitroimidazol-1-yl)-propan-1-ol) have been studied. Crystals are triclinic in the temperature interval between 100 and 333 K (melting point) with two different molecular conformations present in the asymmetric unit (Z' = 2) and two of
Marta Dolores Mudry et al.
Mutation research, 721(1), 108-113 (2011-01-25)
The genotoxicity of two nitroimidazole derivatives, ornidazole (ONZ) and metronidazole (MTZ) in the peripheral blood lymphocytes of Cebus libidinosus (CLI) (Primates, Cebidae) was assessed. Endpoints measured included sister chromatid exchange (SCE) frequency, cell proliferation kinetics (CPK), replication index (RI), mitotic
Haozhe Tang et al.
BMC oral health, 17(1), 163-163 (2017-12-30)
We investigated the clinical and radiographic outcomes of autotransplanted teeth over a follow-up period of 2 to 8 years, and summarize the findings of other relevant studies with regard to the primary factors that influence a good prognosis in such patients.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service