Skip to Content
Merck
All Photos(1)

Key Documents

O4630

Supelco

Orphenadrine citrate salt

analytical standard

Synonym(s):

β-Dimethylaminoethyl 2-methylbenzhydryl ether citrate salt, o-Methyldiphenhydramine citrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23NO · C6H8O7
CAS Number:
Molecular Weight:
461.50
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CC(CC(O)=O)C(O)=O.CN(C)CCOC(c1ccccc1)c2ccccc2C

InChI

1S/C18H23NO.C6H8O6/c1-15-9-7-8-12-17(15)18(20-14-13-19(2)3)16-10-5-4-6-11-16;7-4(8)1-3(6(11)12)2-5(9)10/h4-12,18H,13-14H2,1-3H3;3H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

XLSFUJUURNFSCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Muscarinic receptor antagonist; H1 histamine receptor antagonist; muscle relaxant. Orphenadrine has also been reported to inhibit the noradrenergic transporter and to block the NMDA receptor ion channel.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Saracino et al.
Journal of pharmaceutical and biomedical analysis, 50(3), 501-506 (2009-06-16)
Orphenadrine is an antimuscarinic agent mainly used for the treatment of parkinsonism and to alleviate the neuroleptic syndrome induced by antipsychotic drugs. A new, rapid analytical method, based on liquid chromatography with diode array detection (DAD), has been developed and
Mei-juan Xu et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 42(7), 730-734 (2007-09-22)
To study the enzyme kinetics of schizandrin metabolism in different gender in rat liver microsomes, liver microsomes were prepared from male or female rats. Schizandrin was incubated with rat liver microsomes. Schizandrin and its metabolites were isolated and identified by
Mirosław Czuczwar et al.
Pharmacological reports : PR, 61(4), 732-736 (2009-10-10)
Orphenadrine is an anticholinergic drug used in the treatment of Parkinson's disease, and is also known to exert nonspecific antagonistic activity at the phencyclidine binding site of the N-methyl-D-aspartate (NMDA) receptor. The aim of this study was to assess the
Marianna Borsodi et al.
Orvosi hetilap, 149(39), 1847-1852 (2008-09-23)
The authors compared the potency, safety and tolerability of combined infusion containing non-steroid anti-inflammation diclofenac and central muscle relaxant orphenadrine, and those of tramadol HCl, during postoperative pain relief after low and middle category operations. The test was an open
[Interaction of drugs with the primary disease - when medicines exacerbate illnesses].
Anne Taegtmeyer et al.
Praxis, 101(6), 363-370 (2012-03-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service