Skip to Content
Merck
All Photos(1)

Key Documents

O0120000

Ofloxacin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20FN3O4
CAS Number:
Molecular Weight:
361.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Quality Level

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

CC1COc2c(N3CCN(C)CC3)c(F)cc4C(=O)C(=CN1c24)C(O)=O

InChI

1S/C18H20FN3O4/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21/h7-8,10H,3-6,9H2,1-2H3,(H,24,25)

InChI key

GSDSWSVVBLHKDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 50 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G A Gates
The Pediatric infectious disease journal, 20(1), 104-107 (2001-02-15)
To assess the safety of topical agents in the middle ear, animal studies were reviewed. Compared with aminoglycoside-containing preparations, which caused significant loss of hair cells in the basal turn of the cochlea, ofloxacin caused no loss of hair cells
S V Onrust et al.
Drugs, 56(5), 895-928 (1998-11-26)
Ofloxacin is an established fluoroquinolone agent which achieves good concentrations in genitourinary tract tissues and fluids. It has good in vitro activity against most Enterobacteriaceae, Staphylococcus saprophyticus, methicillin-susceptible S. aureus, Neisseria gonorrhoeae, Chlamydia trachomatis and Haemophilus ducreyi, intermediate activity against
K L Simpson et al.
Drugs, 58(3), 509-531 (1999-09-24)
Ofloxacin is a synthetic fluoroquinolone antibacterial agent that inhibits the supercoiling activity of bacterial DNA gyrase, halting DNA replication. Ofloxacin has been formulated as a 0.3% otic solution for the treatment of ear infections. Topical administration of ofloxacin otic solution
J Kawakami et al.
Journal of pharmacokinetics and biopharmaceutics, 22(3), 207-227 (1994-06-01)
We attempted to predict the delivery of ofloxacin (OFLX), a new quinolone antibacterial agent (NQ), into cerebrospinal fluid (CSF) in the human based on the physiological properties and pharmacokinetic parameters of NQs in various animals. Physiological properties for evaluation of
Chee Wee Gan et al.
The Laryngoscope, 123(7), 1770-1777 (2013-05-15)
To develop a novel drug-eluting biodegradable ventilation tube (VT), to evaluate in vitro sustained release and antibacterial adherence of ofloxacin-loaded biodegradable VT on Pseudomonas aeruginosa, and to evaluate in vivo biodegradation of VT in guinea pig ears. A randomized animal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service