Skip to Content
Merck
All Photos(2)

Key Documents

M6752

Sigma-Aldrich

Methyl salicylate

ReagentPlus®, ≥99% (GC)

Synonym(s):

2-Hydroxybenzoic acid methyl ester, Methyl 2-hydroxybenzoate, Oil of wintergreen, Wintergreen oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
971516
EC Number:
MDL number:
UNSPSC Code:
12191503
PubChem Substance ID:
NACRES:
NA.25

vapor density

5.26 (vs air)

Quality Level

vapor pressure

1 mmHg ( 54 °C)

product line

ReagentPlus®

Assay

≥99% (GC)

form

liquid

autoignition temp.

847 °F

refractive index

n20/D 1.536 (lit.)

bp

222 °C (lit.)

mp

−8-−7 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

COC(=O)c1ccccc1O

InChI

1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3

InChI key

OSWPMRLSEDHDFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl salicylate (MeSA) is a natural herbivore-induced plant volatile It is a naturally occurring product in trees, legumes, exotic plants, vegetables, berries, and the primary constituent of the oil of wintergreen. MeSA is produced from salicylic acid.

Application

Methyl salicylate has been used:
  • as a component of clarifying solution for treating Mongolian gerbil cochlea intact for immunofluorescence analysis
  • as a plant elicitor to test its effect on reducing the whitefly population from tomato plants
  • in the hyalinization of tissues for pulmonary angiography
  • in clearing tissues

Biochem/physiol Actions

Methyl salicylate plays an important role in fruit ripening. It is known to attract natural enemies of herbivores. MeSA inhibits the activity of aminocyclopropane-1-carboxylic acid synthase (ACC synthase) and aminocyclopropane-1-carboxylic acid oxidase (ACC oxidase) in plums and tomatoes, it can inhibit fungal infections and reduce chilling injury symptoms in fruits like pomegranates. This oil of wintergreen is of great interest in the tobacco industry as a flavorant. It has counter irritant and anti-inflammatory effects.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Excitatory postrhinal projections to principal cells in the medial entorhinal cortex
Koganezawa N, et al.
The Journal of Neuroscience, 35(48), 15860-15874 (2015)
Therapeutic Medications in Athletic Training (2007)
Aspirin and the Salicylates (2013)
A novel mouse model of high flow-induced pulmonary hypertension-surgically induced by right pulmonary artery ligation
Zhang A, et al.
The Journal of Surgical Research, 208, 132-139 (2017)
Active Ingredients Used in Cosmetics: Safety Survey (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service