Skip to Content
Merck
All Photos(1)

Key Documents

M5626

Supelco

17α-Methylandrostan-17β-ol-3-one

analytical standard

Synonym(s):

17α-Methyldihydrotestosterone, 17β-Hydroxy-17α-methyl-5α-androstan-3-one, 5α-Androstane-17α-methyl-17β-ol-3-one, Mestaline, Mestanolone, Methylandrostanolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O2
CAS Number:
Molecular Weight:
304.47
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C20H32O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h13,15-17,22H,4-12H2,1-3H3/t13-,15+,16-,17-,18-,19-,20-/m0/s1

InChI key

WYZDXEKUWRCKOB-YDSAWKJFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Gragera et al.
Histology and histopathology, 8(3), 449-455 (1993-07-01)
The effects of anabolic steroid treatment in association with endurance training on biochemical serum parameters and liver ultrastructure have been investigated in male rats. Values of serum alanine aminotransferase, aspartate aminotransferase and alkaline phosphatase were not significantly affected by administration
M S Fitzpatrick et al.
General and comparative endocrinology, 95(3), 399-408 (1994-09-01)
Androgen receptors were identified in the cytosol from ovaries of juvenile coho salmon, Oncorhynchus kisutch. The binding for the synthetic androgen mibolerone was specific and saturable (Kd = 0.32 +/- 0.02 nM; Bmax = 15.31 +/- 4.31 fmol/mg protein). Bound
M E Stobaugh et al.
Steroids, 55(6), 259-262 (1990-06-01)
Dihydrotestosterone-succinyl-agarose is the most common form of androgen affinity column. To investigate the effect of variation in the number of methylene groups between the ester and amide functions, a homologous series, varying the number of methylenes between the functional groups
Masayuki Yamada et al.
Journal of pharmaceutical and biomedical analysis, 45(1), 125-133 (2007-08-22)
Anabolic steroids with the 17alpha-methyl,17beta-hydroxyl group, which were developed as oral formulations for therapeutic purposes, have been abused in the field of human sports. These anabolic steroids are also used to enhance racing performance in racehorses. In humans, structurally related
Frauke Hoffmann et al.
Chemosphere, 87(11), 1246-1253 (2012-02-22)
Endocrine disrupting compounds (EDCs) are well known to interfere with the hormone system of aquatic vertebrates and to affect their reproductive biology. 17α-Methyldihydrotestosterone (MDHT) is a widely used model compound for the assessment of androgenic EDCs, because it binds with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service