Skip to Content
Merck
All Photos(1)

Key Documents

A7755

Sigma-Aldrich

5α-Androstane-3α,17β-diol

Synonym(s):

3α,17β-Dihydroxy-5α-androstane, Dihydroandrosterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H32O2
CAS Number:
Molecular Weight:
292.46
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

Quality Level

form

powder

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

toxicology assay: suitable

solubility

methanol: 19.60-20.40 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C19H32O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-17,20-21H,3-11H2,1-2H3/t12-,13+,14-,15-,16-,17-,18-,19-/m0/s1

InChI key

CBMYJHIOYJEBSB-KHOSGYARSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application

5α-Androstane-3α,17β-diol (dihydroandrosterone) is a testosterone metabolite. Dihyroandrosterone inhibited the cell growth of seven cancer cell lines while showing weak toxicity on normal cell lines.

Biochem/physiol Actions

Dihydroandrosterone is a testosterone metabolite; affects sperm maturation and survival.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheryl A Frye
Pharmacology, biochemistry, and behavior, 86(2), 354-367 (2006-11-23)
The abuse of anabolic-androgenic steroids (AS) is a growing problem; however, the effects and mechanisms underlying their addictive effects are not well understood. Research findings regarding androgen abuse in people and hedonic effects of androgens in laboratory rats are reviewed.
D H Garnier et al.
General and comparative endocrinology, 116(2), 281-290 (1999-11-24)
Concentrations of testosterone, progesterone, Delta4-androstenedione, dihydrotestosterone, 11-ketotestosterone, estrone, estradiol-17beta, 5alpha-androstane, 3alpha, 17beta-diol, and 17alpha-hydroxy, 20beta-dihydroprogesterone were determined by radioimmunoassays in the blood plasma and testicular homogenates of Scyliorhinus canicula. Samples were collected almost every month for 27 months. The weights
Wei Liu et al.
Journal of chromatography. A, 1233, 1-7 (2012-03-06)
In this paper, a convenient and self-assembled hollow fiber solvent-stir bar microextraction (HF-SSBME) device was developed, which could stir by itself. In the extraction process, the proposed device made the solvent "bar" not floating at the sample solution and exposing
J P Deslypere et al.
The Journal of clinical endocrinology and metabolism, 53(2), 430-434 (1981-08-01)
Testosterone, 5 alpha-androstane-17 beta-ol-3-one (DHT) and 5 alpha-androstane-3 alpha,17 beta-diol (Adiol) concentrations were measured in postmortem tissues (pubic skin, scrotal skin, thigh skin, and striated muscle) from 24 males, aged 20-82 yr. Androgen concentrations were highest in scrotal skin, followed
Hajer Jegham et al.
Anti-cancer drugs, 23(8), 803-814 (2012-03-01)
This study investigated the antineoplasic potential of a new family of aminosteroids. The antiproliferative activity of seven 5α-androstane-3α,17β-diol derivatives selected from a screening study was measured on nine cancerous cell lines (HL-60, K-562, LNCaP, PC-3, Shionogi, MCF-7, MDA-MB-231, BT-20, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service