Skip to Content
Merck
All Photos(5)

Key Documents

I8507

Sigma-Aldrich

Iodomethane

Iodomethane

contains copper as stabilizer, ReagentPlus®, 99%

Synonym(s):

Methyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94
Beilstein:
969135
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
41-43 °C (lit.)
vapor pressure:
24.09 psi ( 55 °C)
7.89 psi ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor density

4.89 (vs air)

vapor pressure

24.09 psi ( 55 °C)
7.89 psi ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

contains

copper as stabilizer

dilution

(for general lab use)

refractive index

n20/D 1.531 (lit.)

pH

6.3 (21.1 °C)

bp

41-43 °C (lit.)

mp

−64 (lit.)

density

2.28 g/mL at 25 °C (lit.)

functional group

alkyl halide
iodo

storage temp.

2-8°C

SMILES string

CI

InChI

1S/CH3I/c1-2/h1H3

InChI key

INQOMBQAUSQDDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iodomethane also known as methyl iodide is a methylating agent. Its potential as an alternative to methyl bromide for soil fumigation has been studied.[1] The mechanism of the gas-phase reaction of iodomethane with chlorine atom has been described.[2] The dynamics of photolysis of iodomethane have been studied using multiphoton ionization.[3] The interaction of iodomethane with Cu(111) surface has been analyzed.[4]

Application

Iodomethane (Methyl iodide) has been used to generate iodide water cluster reagent ions for ToF-CIMS (time-of-flight chemical ionization mass spectrometry) analysis.[5] It may be used to prepare (E)-1-iodopropene[6] and N,N,N-trimethylchitosan (TMC).[7]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.8 °F - closed cup

Flash Point(C)

32.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

High-resolution chemical ionization mass spectrometry (ToF-CIMS): application to study SOA composition and processing.
Aljawhary D, et al.
Atmospheric Measurement Techniques, 6(11), 3211-3224 (2013)
Methyl iodide, an ozone-safe alternative to methyl bromide as a soil fumigant.
Ohr HD, et al.
Plant Disease, 80(7), 731-735 (1996)
Methyl rotation, vibration, and alignment from a multiphoton ionization study of the 266 nm photodissociation of methyl iodide.
Loo RO, et al.
J. Chem. Phys. , 90(8), 4222-4236 (1989)
Iodomethane dissociation on Cu (111): Bonding and chemistry of adsorbed methyl groups.
Lin JL and Bent BE.
Journal of Vacuum Science & Technology. A, Vacuum, Surfaces, And Films, 10(4), 2202-2209 (1992)
Methylation of chitosan with iodomethane: effect of reaction conditions on chemoselectivity and degree of substitution.
Curti E, et al.
Macromolecular Bioscience, 3(10), 571-576 (2003)

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the density of the product?

    1 answer
    1. As mentioned in the PROPERTIES section, the density of this product is 2.28 g/mL at 25 °C (lit.).

      Helpful?

  4. What is the density of the product?

    1 answer
    1. As mentioned in the PROPERTIES section, the density of this product is 2.28 g/mL at 25 °C (lit.).

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service