Skip to Content
Merck

Skip To

D4911

Dextran sulfate sodium salt from Leuconostoc spp.

mol wt 6,500-10,000 by GPC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.21
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacterial (Leuconostoc)

Quality Segment

form

powder

mol wt

6,500-10,000 by GPC

composition

Sulfur Content, 16.0-22.0%

color

white to light yellow

useful pH range

6.0-8 (10 g/L)

solubility

H2O: 100 mg/mL

storage temp.

room temp

General description

Dextran sulfate is a sulfated polysaccharide, which is negatively charged. It is produced by the esterification of dextran with chlorosulphonic acid. Dextran is used to treat hemorrhage, burns, surgery and trauma. It improves blood flow and prevents erythrocyte aggregation. Dextran lowers the risk of thrombosis and postoperative pulmonary emboli. It is a ribonuclease inhibitor. Dextran sulfate exhibits antiviral, anticoagulant and adjuvant properties.

Application

Dextran sulfate sodium salt Leuconostoc spp has been used:
  • as a standard to perform polyacrylamide-gel electrophoresis for carbohydrates (C-PAGE)(59) and agarose gel electrophoresis(60)
  • for RNA sequential probing of targets (SPOTs) imaging(61)
  • as a component of 1% injection buffer(62)

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D6924D600152423
biological source

bacterial (Leuconostoc)

biological source

bacterial (Leuconostoc mesenteroides)

biological source

bacterial (Leuconostoc mesenteroides)

biological source

(Leuconostoc spp)

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

solubility

H2O: 100 mg/mL

solubility

H2O: 100 mg/mL

solubility

H2O: 100 mg/mL

solubility

H2O: 100 mg/mL, clear, colorless to faintly greenish-yellow

form

powder

form

powder

form

powder

form

powder

storage temp.

room temp

storage temp.

room temp

storage temp.

-

storage temp.

room temp

color

white to light yellow

color

white to off-white

color

white to off-white

color

white to off-white


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters