Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dextran sulfate sodium salt from
Leuconostoc
spp.
CAS Number:
9011-18-1
Products
Product Category
carbohydrates (8)
dextrans (5)
Brand
Sigma-Aldrich (8)
Biological Source
Color
Feature
Manufacturer
Physical Form
Research Area
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
D8906
Dextran sulfate sodium salt from
Leuconostoc
spp.,
for molecular biology, average M
w
>500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Expand
View Pricing
31404
Dextran sulfate sodium salt from
Leuconostoc
spp.,
M
r
5,000
Expand
View Pricing
31395
Dextran sulfate sodium salt from
Leuconostoc
spp.,
M
r
~500,000
Expand
View Pricing
D6001
Dextran sulfate sodium salt from
Leuconostoc
spp.,
average mol wt >500,000 (dextran starting material), contains 0.5-2.0% phosphate buffer, pH 6-8
Expand
View Pricing
D4911
Dextran sulfate sodium salt from
Leuconostoc
spp.,
mol wt 6,500-10,000
Expand
View Pricing
D6924
Dextran sulfate sodium salt from
Leuconostoc
spp.,
average mol wt 9,000-20,000
Expand
View Pricing
52423
Dextran sulfate sodium salt from
Leuconostoc
spp.,
low sulfate content, M
r
~40,000
Expand
View Pricing
D8787
Dextran sulfate sodium salt from
Leuconostoc
spp.,
avg mol wt >500,000 (dextran starting material)
Expand
View Pricing