Skip to Content
Merck
All Photos(1)

Key Documents

D0720800

Dexchlorpheniramine maleate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

S-(+)-Chlorpheniramine maleate salt, (S)-γ-(4-Chlorophenyl)-N,N-dimethyl-2-pyridinepropanamine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClN2 · C4H4O4
CAS Number:
Molecular Weight:
390.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dexchlorpheniramine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.CN(C)CC[C@@H](c1ccc(Cl)cc1)c2ccccn2

InChI

1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t15-;/m0./s1

InChI key

DBAKFASWICGISY-DASCVMRKSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dexchlorpheniramine maleate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anaphylactic reaction to ranitidine and dexchlorpheniramine.
C Thurot-Guillou et al.
European journal of dermatology : EJD, 17(2), 170-171 (2007-03-06)
Urticaria due to aloe vera: a new sensitizer?
Alberto Alvarez-Perea et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 105(5), 404-405 (2010-11-09)
O Cáceres Calle et al.
Allergologia et immunopathologia, 32(5), 306-309 (2004-10-01)
Dexchlorpheniramine (DH) is a classical or first generation antihistamine belonging to the ethanolamine group. Adverse effects related to these antihistamines are frequent, but the hypersensitivity reactions described in the literature since 1940 are exceptional. We report the case of a
Eef L Theunissen et al.
British journal of clinical pharmacology, 61(1), 79-86 (2006-01-05)
Previous studies have demonstrated that the antihistamines mequitazine, cetirizine and dexchlorpheniramine produce mild sedation after single doses. It is unknown, however, whether acute sedation persists after repeated dosing. Therefore, this study assessed the effects of repeated dosing of these antihistamines
José R Villada et al.
Journal of cataract and refractive surgery, 31(3), 620-621 (2005-04-07)
We report a severe anaphylactic reaction that occurred about 5 minutes after 1.0 mg of cefuroxime was injected into the anterior chamber after routine phacoemulsification and intraocular lens implantation. The patient was known to be allergic to penicillin. Immediate action

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service