Skip to Content
Merck
All Photos(1)

Key Documents

C2600000

Colistimethate sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Colistin sodium methanesulfonate, Colimycin sodium methanesulfonate, Methanesulfonic acid derivative of Polymyxin E, Polymyxin E sodium methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H115N16Na5O28S5
CAS Number:
Molecular Weight:
1759.90
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

colistimethate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].CS([O-])(=O)=O.CS([O-])(=O)=O.CS([O-])(=O)=O.CS([O-])(=O)=O.CS([O-])(=O)=O.CCC(C)CCCCC(=O)N[C@@H](CCN)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](CCN)C(=O)N[C@H]1CCNC(=O)[C@@H](NC(=O)[C@H](CCN)NC(=O)[C@@H](CCN)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](CC(C)C)NC(=O)[C@H](CCN)NC1=O)C(C)O

InChI

1S/C53H100N16O13.5CH4O3S.5Na/c1-9-30(6)12-10-11-13-41(72)60-33(14-20-54)48(77)69-43(32(8)71)53(82)65-36(17-23-57)45(74)64-38-19-25-59-52(81)42(31(7)70)68-49(78)37(18-24-58)62-44(73)34(15-21-55)63-50(79)39(26-28(2)3)67-51(80)40(27-29(4)5)66-46(75)35(16-22-56)61-47(38)76;5*1-5(2,3)4;;;;;/h28-40,42-43,70-71H,9-27,54-58H2,1-8H3,(H,59,81)(H,60,72)(H,61,76)(H,62,73)(H,63,79)(H,64,74)(H,65,82)(H,66,75)(H,67,80)(H,68,78)(H,69,77);5*1H3,(H,2,3,4);;;;;/q;;;;;;5*+1/p-5/t30?,31-,32-,33+,34-,35+,36+,37+,38+,39+,40-,42+,43+;;;;;;;;;;/m1........../s1

InChI key

WSDSONZFNWDYGZ-BKFDUBCBSA-I

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Colistimethate sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Mode of Action: Binds to lipids on the cell cytoplasmic membrane of Gram-negative bacteria and disrupts the cell wall integrity.
Antimicrobial spectrum: Gram-negative bacteria.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S M Garonzik et al.
Antimicrobial agents and chemotherapy, 55(7), 3284-3294 (2011-05-11)
With increasing clinical emergence of multidrug-resistant Gram-negative pathogens and the paucity of new agents to combat these infections, colistin (administered as its inactive prodrug colistin methanesulfonate [CMS]) has reemerged as a treatment option, especially for critically ill patients. There has
Kavi J Littlewood et al.
Journal of cystic fibrosis : official journal of the European Cystic Fibrosis Society, 11(5), 419-426 (2012-06-22)
Various inhaled antibiotics are currently used for treating chronic Pseudomonas aeruginosa lung infection in cystic fibrosis (CF) patients, however their relative efficacies are unclear. We compared the efficacy of the inhaled antibiotics tobramycin (TIP, TIS-T, TIS-B), colistimethate sodium (colistin) and
W Couet et al.
Clinical pharmacology and therapeutics, 89(6), 875-879 (2011-05-06)
Colistin pharmacokinetics (PK) was investigated in young healthy volunteers after a 1-h infusion of 80 mg (1 million international units (MIU)) of the prodrug colistin methanesulfonate (CMS). Concentration levels of CMS and colistin were determined in plasma and urine using
Stephanie J Wallace et al.
Journal of pharmaceutical sciences, 101(9), 3347-3359 (2012-05-25)
Interaction of colistin and colistin methanesulfonate (CMS) with liposomes has been studied with the view to understanding the limitations to the use of liposomes as a more effective delivery system for pulmonary inhalation of this important class of antibiotic. Thus
Matti Karvanen et al.
Antimicrobial agents and chemotherapy, 57(1), 668-671 (2012-11-14)
This report describes the pharmacokinetics of colistin methanesulfonate (CMS) and colistin in five intensive care unit patients receiving continuous venovenous hemodiafiltration. For CMS, the mean maximum concentration of drug in plasma (C(max)) after the fourth dose was 6.92 mg/liter and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service