Skip to Content
Merck

Skip To

BP995

Fluorouracil

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

5-Fluorouracil, 2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C4H3FN2O2
CAS Number:
Molecular Weight:
130.08
EC Number:
200-085-6
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
127172
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

fluorouracil

form

solid

quality

British Pharmacopoeia (BP) Reference Standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

282-286 °C (dec.) (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

FC1=CNC(=O)NC1=O

InChI

1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

GHASVSINZRGABV-UHFFFAOYSA-N

Gene Information

human ... TYMS(7298)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluorouracil BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 40 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
03738F6627F0250000
grade

pharmaceutical primary standard

grade

analytical standard

grade

-

grade

pharmaceutical primary standard

manufacturer/tradename

BP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

EDQM

form

solid

form

-

form

powder

form

-

format

neat

format

neat

format

-

format

neat

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

cleaning products
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

pharmaceutical (small molecule)

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under Fluorouracil


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters