Skip to Content
Merck
All Photos(1)

Documents

BP576

Hydrocortisone

British Pharmacopoeia (BP) Assay Standard

Synonym(s):

11β,17α,21-Trihydroxypregn-4-ene-3,20-dione, 17-Hydroxycorticosterone, 4-Pregnene-11β,17α,21-triol-3,20-dione, Cortisol, Kendall’s compound F, Reichstein’s substance M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O5
CAS Number:
Molecular Weight:
362.46
Beilstein:
1354819
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

hydrocortisone

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

211-214 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

JYGXADMDTFJGBT-VWUMJDOOSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydrocortisone BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Tretinoin, Hydrocortisone and Hydroquinone Cream
  • Miconazole and Hydrocortisone Acetate Cream
  • Hydrocortisone Sodium Succinate Injection
  • Prednisolone Tablets
  • Miconazole and Hydrocortisone Cream
  • Miconazole and Hydrocortisone Ointment
  • Hydrocortisone and Neomycin Cream
  • Hydrocortisone and Clioquinol Ointment
  • Hydrocortisone Sodium Phosphate Oral Solution
  • Prednisolone Gastro-resistant Tablets
  • Hydrocortisone Sodium Phosphate
  • Hydrocortisone Sodium Phosphate Injection
  • Hydrocortisone and Clioquinol Cream
  • Methylprednisolone Tablets
  • Hydrocortisone Acetate Oral Suspension
  • Hydrocortisone Cream
  • Hydrocortisone Oromucosal Tablets
  • Biochem/physiol Actions

    Primary glucocorticoid secreted by the adrenal cortex. It has three times the anti-inflammatory potency of corticosterone but much lower Na2+ retention potency.

    Other Notes

    Sales restrictions may apply.

    related product

    Product No.
    Description
    Pricing

    Pictograms

    Health hazard

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Repr. 1A - STOT RE 2

    Target Organs

    Adrenal gland,Bone marrow

    Storage Class Code

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    WGK

    WGK 2

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Karin E Koopman et al.
    Hepatology (Baltimore, Md.), 60(2), 545-553 (2014-03-29)
    American children consume up to 27% of calories from high-fat and high-sugar snacks. Both sugar and fat consumption have been implicated as a cause of hepatic steatosis and obesity but the effect of meal pattern is largely understudied. We hypothesized
    Liselotte van Bloemendaal et al.
    Diabetes, 63(12), 4186-4196 (2014-07-30)
    Gut-derived hormones, such as GLP-1, have been proposed to relay information to the brain to regulate appetite. GLP-1 receptor agonists, currently used for the treatment of type 2 diabetes (T2DM), improve glycemic control and stimulate satiety, leading to decreases in
    Mario Cioce et al.
    Oncotarget, 5(12), 4129-4143 (2014-07-02)
    Metabolic remodeling is a hallmark of cancer progression and may affect tumor chemoresistance. Here we investigated by 1H-NMR/PCA analysis the metabolic profile of chemoresistant breast cancer cell subpopulations (ALDHbright cells) and their response to metformin, a promising anticancer metabolic modulator.
    Florian Hardeveld et al.
    Psychoneuroendocrinology, 50, 62-71 (2014-09-03)
    Although HPA-axis activity has been studied extensively in relation to depression, there is no consensus whether HPA-axis parameters predicts major depressive disorder (MDD) recurrence. We investigated whether HPA-axis parameters (cortisol awakening response (CAR), the dexamethasone suppression test (DST) and evening
    Lorenz Goette et al.
    Psychoneuroendocrinology, 54, 115-123 (2015-02-24)
    Social competition is a fundamental mechanism of evolution and plays a central role in structuring individual interactions and communities. Little is known about the factors that affect individuals' competitive success, particularly in humans. Key factors might include stress, a major

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service