Skip to Content
Merck
All Photos(2)

Key Documents

94377

Supelco

Indeno[1,2,3-cd]pyrene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
Beilstein:
1879312
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

c1ccc-2c(c1)-c3ccc4ccc5cccc6cc-2c3c4c56

InChI

1S/C22H12/c1-2-7-17-16(6-1)18-11-10-14-9-8-13-4-3-5-15-12-19(17)22(18)21(14)20(13)15/h1-12H

InChI key

SXQBHARYMNFBPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J E Rice et al.
Carcinogenesis, 11(11), 1971-1974 (1990-11-01)
Indeno[1,2,3-cd]pyrene (IP) is a non-alternant polycyclic aromatic hydrocarbon that has tumor-initiating activity on mouse skin and is carcinogenic in newborn mice and in rat lungs. Previous studies have shown that 8- and 9-hydroxyIP and IP-1,2-diol are major metabolites formed in
Yi-Hsin Tseng et al.
Ecotoxicology and environmental safety, 210, 111892-111892 (2021-01-12)
Human activities have generated air pollution, with extremely small particles (PM 2.5, particulate matter less than 2.5 µm in diameter) and liquid droplets, which become a menace to human health. Among the pollutants, polycyclic aromatic hydrocarbons (PAHs), which enhance the risks
Luke Chandler Short et al.
Journal of the American Society for Mass Spectrometry, 18(4), 589-599 (2006-12-26)
The technique of atmospheric pressure photoionization (APPI) has several advantages over electrospray ionization (ESI) and atmospheric pressure chemical ionization (APCI), including efficient ionization of nonpolar or low charge affinity compounds, reduced susceptibility to ion suppression, high sensitivity, and large linear
Nitration of indeno[1,2,3-cd]pyrene and mutagenic activities of related compounds.
M Minabe et al.
Chemical research in toxicology, 2(6), 357-358 (1989-11-01)
Indeno [1,2,3-cd]pyrene.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 32, 373-378 (1983-12-01)

Articles

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service