Skip to Content
Merck
All Photos(2)

Documents

93893

Supelco

Physcion

analytical standard

Synonym(s):

1,8-Dihydroxy-3-methoxy-6-methylanthraquinone, 6-O-Methylemodin, 6-Methoxychrysophanic acid, Emodin-3-methyl ether, Parietin, Rheochrysidin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
Beilstein:
6770499
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(O)c2C(=O)c3c(O)cc(C)cc3C(=O)c2c1

InChI

1S/C16H12O5/c1-7-3-9-13(11(17)4-7)16(20)14-10(15(9)19)5-8(21-2)6-12(14)18/h3-6,17-18H,1-2H3

InChI key

FFWOKTFYGVYKIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Physcion is an anthraquinone derivative and is available as the active constituent of Chinese medicinal plants such as Rhubarb, Polygonum cuspidatum and Reynoutria multiflora. It exhibits various biological activities such as antitumor, antimicrobial, antifungal, antioxidant and antihuman cytomegalovirus activity.

Application

Physcion may be used as an analytical reference standard for the quantification of the analyte in medicinal plants, pharmaceutical preparations, vegetables, herbs and liquors using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Anthraquinone-derivative found in medicinal herbs.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Occurrence of emodin, chrysophanol and physcion in vegetables, herbs and liquors. Genotoxicity and anti-genotoxicity of the anthraquinones and of the whole plants
Mueller O.S, et al.
Food And Chemical Toxicology, 37(5), 481-491 (1999)
Ming-Yi Shen et al.
PloS one, 6(11), e27480-e27480 (2011-11-17)
Plants provide a rich source of lead compounds for a variety of diseases. A novel approach combining phytochemistry and chemotaxis assays was developed and used to identify and study the mechanisms of action of the active compounds in F. japonica
Simultaneous determination of five anthraquinones in medicinal plants and pharmaceutical preparations by HPLC with fluorescence detection
He D, et al.
Journal of Pharmaceutical and Biomedical Analysis, 49(4), 1123-1127 (2009)
Han-Jing Yan et al.
The American journal of Chinese medicine, 38(3), 473-483 (2010-05-27)
Fallopia multiflora (Thunb.) Harald, a traditional Chinese medicinal plant, is used in treating dizziness. In this study, the samples of F. multiflora from ten different locations were collected, and five bioactive components (2, 3, 5, 4'-tetrahydroxystilbene-2-O-beta-D-glucoside, emodin, emodin-8-O-beta-D-glucoside, physcion and
Santosh Pandit et al.
Biofouling, 28(3), 279-287 (2012-03-23)
Polygonum cuspidatum is a plant with spreading rhizomes and numerous reddish-brown stems that has been used in Korean folk medicine to improve oral hygiene. Nevertheless, there are no reports related to its possible effect on the virulence of dental biofilms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service