Skip to Content
Merck
All Photos(2)

Key Documents

15721

Sigma-Aldrich

Polydatin

≥95% (HPLC)

Synonym(s):

Piceid, Reservatrol-3-β-mono-D-glucoside, Resveratrol 3-O-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22O8
CAS Number:
Molecular Weight:
390.38
Beilstein:
54837
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (HPLC)

form

powder

optical activity

[α]1/D -66.0±2.0° in ethanol

composition

carbon, 60.5-62.5%

storage temp.

2-8°C

SMILES string

OCC1O[C@@H](Oc2cc(O)cc(\C=C\c3ccc(O)cc3)c2)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C20H22O8/c21-10-16-17(24)18(25)19(26)20(28-16)27-15-8-12(7-14(23)9-15)2-1-11-3-5-13(22)6-4-11/h1-9,16-26H,10H2/b2-1+/t16?,17-,18+,19-,20-/m1/s1

InChI key

HSTZMXCBWJGKHG-AEGFYQLOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polydatin, a type of polyphenolic phytoalexin, has many physiological and pharmacological effects including anti-inflammatory and anti-oxidative activities. Polydatin is an effective candidate drug for the protection of photo-inflammation. Polydatin exhibits therapeutic potential for vascular dementia, most likely due to its anti-oxidant activity and the direct protection of neurons.

Biochem/physiol Actions

Stilbene found in medicinal herbs. Antioxidant, anti-inflammatory and neuroprotective.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Zhang et al.
PloS one, 7(9), e46574-e46574 (2012-10-03)
Polydatin is one of main compounds in Polygonum cuspidatum, a plant with both medicinal and nutritional value. The possible hepatoprotective effects of polydatin on acute liver injury mice induced by carbon tetrachloride (CCl(4)) and the mechanisms involved were investigated. Intraperitoneal
Maria Rotches-Ribalta et al.
Pharmacological research, 66(5), 375-382 (2012-08-22)
A pharmacokinetic study of the metabolic profile of resveratrol has been performed in healthy men after moderate red wine (RW) consumption. The bioavailability of resveratrol is highly influenced by several factors such as the food matrix and, therefore, this study
Xingmin Wang et al.
Expert opinion on investigational drugs, 22(2), 169-179 (2012-12-18)
The aim of the study was find out whether neuronal mitochondrial injury does take place in severe shock and to explore effective therapy for severe shock. Rats were divided in the following group: sham, shock + normal saline (NS), shock
Giampietro Ravagnan et al.
Inflammation, 36(1), 26-34 (2012-09-08)
It is well known that human keratinocytes produce the anti-microbial peptide β-defensin 2. Its production is enhanced by pathogenic microorganisms or other environmental stressors. In this study, we evaluated the effect of resveratrol, a polyphenol found in several dietary source
Hui Wen et al.
Biomedical microdevices, 14(4), 721-728 (2012-04-25)
Caenorhabditis elegans (C. elegans) is an excellent model organism for the study of aging and longevity. In this work, we presented a microfluidic approach for the evaluation of longevity in C. elegans under stress. The microfluidic device integrated multiple microvalves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service