Skip to Content
Merck
All Photos(1)

Documents

525197

Supelco

Bis(2-ethylhexyl) adipate

99%

Synonym(s):

Adipic acid di(2-ethylhexyl) ester, DOA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH2CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
370.57
Beilstein:
1803774
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

Assay

99%

refractive index

n20/D 1.447 (lit.)

bp

175 °C/2 mmHg (lit.)

mp

<−70 °C (lit.)

density

0.925 g/mL at 20 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

InChI

1S/C22H42O4/c1-5-9-13-19(7-3)17-25-21(23)15-11-12-16-22(24)26-18-20(8-4)14-10-6-2/h19-20H,5-18H2,1-4H3

InChI key

SAOKZLXYCUGLFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2-ethylhexyl) adipate (Bis(2-ethylhexyl) phthalate (DEHP)), is an ester of adipic acid and 2-ethylhexanol. It is a plasticizer and finds application in plastic technology, in order to impart flexibility to rigid polymers. It is an indirect food additive formed due to contact of adhesives with polymers.

Application

Plasticizer

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oral toxicity of bis (2-ethylhexyl) phthalate during pregnancy and suckling in the Long?Evans rat.
Food And Chemical Toxicology, 36 (11), 963-970 (1998)
Yadollah Yamini et al.
Journal of separation science, 32(18), 3201-3208 (2009-09-12)
Two approaches based on solidification of floating drop microextraction (SFDME) and homogenous liquid-liquid microextraction (HLLE) were compared for the extraction and preconcentration of di-(2-ethylhexyl) phthalate (DEHP) and di-(2-ethylhexyl) adipate (DEHA) from the mineral water samples. In SFDME, a floated drop
Julie Borch et al.
Reproductive toxicology (Elmsford, N.Y.), 19(4), 517-525 (2005-03-08)
This study aimed to characterize the effects of di(2-ethylhexyl) phthalate (DEHP) on the fetal rat testes and relate them to the effects seen in adults. Histopathological effects in fetal testes were examined with immunohistochemistry for anti-Mullerian hormone (AMH), 3beta-hydroxysteroid dehydrogenase
Stéphane C Fayoux et al.
Journal of food science, 72(4), E143-E154 (2007-11-13)
In order to better understand and optimize the sorption of limonin (the major navel orange juice bitter principle) by various plasticized polymeric films, a sorption and plasticizer migration study was carried out using high-performance liquid chromatography (HPLC) and gas chromatography
Kirsten Jarfelt et al.
Reproductive toxicology (Elmsford, N.Y.), 19(4), 505-515 (2005-03-08)
Di(2-ethylhexyl) phthalate (DEHP) is a well-known testicular toxicant inducing adverse effects in androgen responsive tissues. Therefore, di(2-ethylhexyl) adipate (DEHA) is currently being evaluated as a potential substitute for DEHP. Similarities in structure and metabolism of DEHP and DEHA have led

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service