Skip to Content
Merck

92142

Biochanin A

analytical standard

Synonym(s):

4′-Methylgenistein, 5,7-Dihydroxy-4′-methoxyisoflavone, Genistein 4′-methyl ether, Olmelin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
EC Number:
207-744-7
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
278107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

210-213 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

COc1ccc(cc1)C2=COc3cc(O)cc(O)c3C2=O

InChI

1S/C16H12O5/c1-20-11-4-2-9(3-5-11)12-8-21-14-7-10(17)6-13(18)15(14)16(12)19/h2-8,17-18H,1H3

InChI key

WUADCCWRTIWANL-UHFFFAOYSA-N

Biochem/physiol Actions

Biochanin A is an isoflavone phytoestrogen found in red clover (Trifolium pratense) that is a selective agonist at ER-β estrogen receptors, and may have chemopreventive efficacy against breast cancer. In line with its low activity at ER-α estrogen receptors, it is essentially devoid of uterotrophic activity. Biochanin A is also a ligand for the aryl hydrocarbon receptor (AhR). It reduces arterial resistance and enhances microcirculation perhaps via effects on potassium and/or calcium ion channels.
Biochanin A is an isoflavone phytoestrogen found in red clover (Trifolium pratense) that is a selective agonist at ER-β estrogen receptors, and may have chemopreventive efficacy against breast cancer. In line with its low activity at ER-α estrogen receptors, it is essentially devoid of uterotrophic activity. Biochanin A is also a ligand for the aryl hydrocarbon receptor (AhR). It reduces arterial resistance and enhances microcirculation perhaps via effects on potassium and/or calcium ion channels. Induction of sulfotransferases for xenobiotic detoxification has been proposed as a mechanism of its cancer preventive effects.


Still not finding the right product?

Explore all of our products under Biochanin A


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library