Skip to Content
Products
Cart
0
AF
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
Biochanin A
Synonym(s):
4′-Methylgenistein, 5,7-Dihydroxy-4′-methoxyisoflavone, Genistein 4′-methyl ether, Olmelin
Empirical Formula (Hill Notation):
C
16
H
12
O
5
CAS Number:
491-80-5
Molecular Weight:
284.26
EC Number:
207-744-7
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
analytical standard (2)
pharmaceutical primary standard (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
D2016
Biochanin A,
Expand
View Pricing
PHL80012
Biochanin A,
phyproof
®
Reference Substance
Expand
View Pricing
1071381
Biochanin A,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
92142
Biochanin A,
analytical standard
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More