Skip to Content
Merck
All Photos(4)

Documents

901916

Sigma-Aldrich

Sodium sulfite

anhydrous, Redi-Dri, reagent grade, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO3
CAS Number:
Molecular Weight:
126.04
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

grade

anhydrous
reagent grade

Agency

suitable for SM 5210

product line

Redi-Dri

Assay

≥98%

form

powder or crystals

pH

9-10.5 (25 °C, 126 g/L)

SMILES string

[Na+].[Na+].[O-]S([O-])=O

InChI

1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

GEHJYWRUCIMESM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amber R Boles et al.
Biotechnology and bioengineering, 109(3), 637-646 (2011-10-22)
A novel sulfur-utilizing perchlorate reducing bacterial consortium successfully treated perchlorate (ClO₄⁻) in prior batch and bench-scale packed bed reactor (PBR) studies. This study examined the scale up of this process for treatment of water from a ClO ₄⁻ and RDX contaminated
Ben M Goff et al.
Journal of the science of food and agriculture, 92(4), 751-758 (2011-11-19)
A variety of methods have been developed for estimating lignin concentration within plant materials. The objective of this study was to compare the lignin concentrations produced by six methods on a diverse population of forage and biomass materials and to
Fabian Leinisch et al.
Chemical research in toxicology, 24(12), 2217-2226 (2011-10-19)
Nitrone spin traps such as 5,5-dimethyl-1-pyrroline-N-oxide (DMPO) are commonly used for free radical detection. Though proven examples are rare, artifact formation must be considered. For example, the Forrester-Hepburn mechanism yields the same radical adduct as that formed by genuine radical
Jingang Wang et al.
Chemosphere, 91(3), 351-357 (2012-12-26)
This paper presents a simple, easy and reliable method for determination of EDTA (ethylenediaminetetraacetic acid) in aqueous system. Using EDTA chelating with ferric irons, an excessive amount of Fe(3+) was added to EDTA solution, and Fe(3+) in excess was reduced
Michelle Cilia et al.
PloS one, 7(10), e48177-e48177 (2012-11-03)
Circulative transmission of viruses in the Luteoviridae, such as cereal yellow dwarf virus (CYDV), requires a series of precisely orchestrated interactions between virus, plant, and aphid proteins. Natural selection has favored these viruses to be retained in the phloem to

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service