Skip to Content
Merck
All Photos(1)

Key Documents

08981

Supelco

Sodium sulfite

analytical standard

Synonym(s):

E221

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO3
CAS Number:
Molecular Weight:
126.04
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98% (RT)

shelf life

limited shelf life, expiry date on the label

solubility

H2O: 1 g/10 mL, clear, colorless

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].[O-]S([O-])=O

InChI

1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

GEHJYWRUCIMESM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application


  • Oxygen scavenging and contaminant removal: Sodium sulfite′s efficacy in removing organic contaminants through oxygen scavenging in various chemical systems was documented, illustrating its critical role in environmental cleanup and wastewater treatment technologies (Zhou et al., 2023).

  • Disinfection by-product management: Sodium sulfite is reviewed for its impact and mechanisms as a residual chlorine quencher, highlighting its potential to reduce harmful disinfection by-products in water treatment processes, which is vital for ensuring safe pharmaceutical and biotechnical operations (Li et al., 2023).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hazem Matar et al.
Cutaneous and ocular toxicology, 36(3), 211-219 (2016-11-08)
Context Incontinence-associated dermatitis (IAD) is a type of moisture-associated dermatitis caused by repeated skin exposure to urine or stool. A product that could mitigate such symptoms would have a significant impact on cost of care and patients' quality of life.
J-S Eun et al.
Journal of dairy science, 97(12), 7833-7843 (2014-09-30)
This experiment was performed to test a hypothesis that nutritive benefits of feeding high-moisture corn (HMC) would be different when fed with different qualities of alfalfa hay (AH) due to associative effects on ruminal fermentation and nutrient utilization efficiency. Eight
Květoslava Křížková et al.
PloS one, 9(11), e112883-e112883 (2014-11-26)
The insulin gene mutation c.137G>A (R46Q), which changes an arginine at the B22 position of the mature hormone to glutamine, causes the monogenic diabetes variant maturity-onset diabetes of the young (MODY). In MODY patients, this mutation is heterozygous, and both
Picheng Gao et al.
Dalton transactions (Cambridge, England : 2003), 44(2), 773-781 (2014-11-20)
A facile synthetic route for TiO2-CdSe heterostructures was proposed based on dentate binding of TiO2 to carboxyl. Carboxyl functionalized CdSe quantum dots (CF-CdSe QDs) were successfully bonded onto TiO2 nanoparticles (NPs), which could significantly improve the photoelectrochemical (PEC) properties of
H A Ramirez Ramirez et al.
Journal of dairy science, 98(3), 1903-1914 (2014-12-31)
Two experiments were conducted to evaluate the additive effects of starch and fat as risk factors associated with milk fat depression in dairy diets containing corn dried distillers grains with solubles. In experiment 1, 4 multiparous ruminally cannulated Holstein cows

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service