Skip to Content
Merck
All Photos(1)

Key Documents

900645

Sigma-Aldrich

Dimethyl sulfoxide

anhydrous, ZerO2®, ≥99.9%

Synonym(s):

Degassed and low oxygen dimethyl sulfoxide, Dimethyl sulfoxide ZerO2®, DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
EC Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.21
grade:
ZerO2®
Assay:
≥99.9%
bp:
189 °C (lit.)
vapor pressure:
0.42 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ZerO2®

Quality Level

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

573 °F

quality

anhydrous

expl. lim.

42 %, 63 °F

impurities

≤0.005% water
water

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

density

1.10 g/mL (lit.)

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service