Skip to Content
Merck
All Photos(1)

Key Documents

87208

Supelco

Myristyltrimethylammonium bromide

suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)

Synonym(s):

Tetradecyltrimethylammonium bromide, Trimethyl(tetradecyl)ammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13N(Br)(CH3)3
CAS Number:
Molecular Weight:
336.39
Beilstein:
3633227
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21
Pricing and availability is not currently available.

description

cationic

Quality Level

Assay

≥99.0% (AT)

form

crystals

quality

LiChropur

mol wt

micellar avg mol wt 27,000

aggregation number

80

technique(s)

ion pair chromatography: suitable

CMC

4-5 mM (20-25°C)

mp

245-250 °C (lit.)

λ

10 % in H2O

UV absorption

λ: 240 nm Amax: ≤0.08
λ: 250 nm Amax: ≤0.05
λ: 260 nm Amax: ≤0.04
λ: 500 nm Amax: ≤0.02

suitability

corresponds to standard for filter test

SMILES string

[Br-].CCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C17H38N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;/h5-17H2,1-4H3;1H/q+1;/p-1

InChI key

CXRFDZFCGOPDTD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ion-pairing reagents as mobile phase additives facilitate the separation of ionic and highly polar substances on reversed phase HPLC columns. Their purity is an important parameter for their successful application. Sigma-Aldrich offers an wide range of tailor-made reagents for anionic (quaternary ammonium and phosphonium salts) and cationic (alkanesulfonates) analyzes. All mobile phase additives are subject to rigorous testing with special emphasis on the requirements of modern reversed phase HPLC:

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dulan B Gunasekara et al.
Electrophoresis, 32(8), 832-837 (2011-03-26)
The combination of microchip electrophoresis with amperometric detection leads to a number of analytical challenges that are associated with isolating the detector from the high voltages used for the separation. While methods such as end-channel alignment and the use of
Yuri Borodko et al.
Langmuir : the ACS journal of surfaces and colloids, 25(12), 6665-6671 (2009-04-29)
The vibrational spectra of platinum nanoparticles (12 nm) capped with tetradecyltrimethylammonium bromide, C(14)TAB, were investigated by Fourier transform infrared (FTIR) spectroscopy. We have shown that the thermal decay of Pt-C(14)TAB nanoparticles in N(2), H(2), and O(2) atmospheres leads to the
Yan Jiang et al.
The journal of physical chemistry. B, 113(24), 8357-8361 (2009-05-26)
Three kinds of conventional surfactants, namely, two nonionic surfactants [polyethylene glycol (23) lauryl ether (Brij-35) and Triton X-100 (TX-100)], one cationic surfactant [n-tetradecyltrimethyl ammonium bromide (TTAB)], and an anionic surfactant [sodium n-dodecyl sulfate (SDS)}, were mixed into the quaternary ammonium
Aline Delbos et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 84(1 Pt 1), 011404-011404 (2011-08-27)
We investigate experimentally the behavior of liquid foams pumped at a given flow rate through a single pore, in the situation where the pore diameter is smaller than the bubble diameter. Results reveal that foam invasion can be observed only
Hayat M Marafie et al.
Talanta, 91, 26-32 (2012-03-01)
Two conventional plastic membrane electrodes that are selective for the tetradecyltrimethylammonium cation (TTA) have been prepared. The ion exchangers of these sensors were the ion associate, TTA-PT, and the ion aggregate, TTA-PSS, where PT and PSS are phosphotungstate and polystyrene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service