79599
VRE ChromoSelect Agar Base
NutriSelect® Plus, suitable for microbiology
Synonym(s):
ChromoSelect Vancomycine Resistant Enterococci Agar Base
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 15.0 g/L
buffering agent, 1.25 g/L
chromogenic mixture, 0.45 g/L
peptone special, 25.0 g/L
salt mixture, 4.25 g/L
sodium chloride, 5.0 g/L
manufacturer/tradename
NutriSelect® Plus
final pH
6.5±0.2 (25 °C)
application(s)
clinical testing
environmental
food and beverages
veterinary
microbiology
storage temp.
2-8°C
suitability
Enterococcus spp.
Related Categories
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service