Skip to Content
Merck
All Photos(1)

Documents

77869

Supelco

(S)-(−)-α-Methylbenzylamine

for chiral derivatization, LiChropur, ≥99.0%

Synonym(s):

(S)-(−)-1-Phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
2204907
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for chiral derivatization

Quality Level

vapor pressure

0.5 mmHg ( 20 °C)

Assay

≥99.0% (sum of enantiomers, GC)
≥99.0%

form

liquid

optical activity

[α]20/D −30±1°, c = 10% in ethanol

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

quality

LiChropur

technique(s)

HPLC: suitable

refractive index

n20/D 1.526 (lit.)
n20/D 1.528

bp

187 °C (lit.)

density

0.94 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H](N)c1ccccc1

InChI

1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3/t7-/m0/s1

InChI key

RQEUFEKYXDPUSK-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(−)-α-Methylbenzylamine is a chiral derivatizing agent, which is employed for derivatizing enantiomers into diastereoisomers.

Application

(S)-(−)-α-Methylbenzylamine may be used as a chiral derivatizating reagent for the determination of acetyl-D-carnitine (D-AC) in acetyl-L-carnitine (L-AC) using high-performance liquid chromatographic (HPLC) enantioseparation method.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Enantiomeric purity determination of acetyl-L-carnitine by reversed-phase high-performance liquid chromatography using chiral derivatization
Kagawa M, et al.
Journal of Chromatography A, 857(1-2), 127-135 (1999)
Manuel Algarra et al.
Journal of separation science, 33(2), 143-154 (2009-12-31)
Free fatty acids were derivatized as amides (DFFA) by reaction with (R)-(+)-1-phenylethylamine, using a simple, fast and robust reaction scheme. A HPLC method with diode array and ESI MS detection was developed for the analysis of the derivatized substances. Six
Shao-Hua Huang et al.
Magnetic resonance in chemistry : MRC, 47(5), 423-427 (2009-02-19)
Two types of chiral analytes, the urea and amide derivatives of alpha-phenylethylamine, were prepared. The effect of inter-molecular hydrogen-bonding interaction on self-discrimination of the enantiomers of analytes has been investigated using high-resolution (1)H NMR. It was found that the urea
Karim Engelmark Cassimjee et al.
Organic & biomolecular chemistry, 10(28), 5466-5470 (2012-06-13)
For biocatalytic production of pharmaceutically important chiral amines the ω-transaminase enzymes have proven useful. Engineering of these enzymes has to some extent been accomplished by rational design, but mostly by directed evolution. By use of a homology model a key
Yan-Song Zheng et al.
Chemical communications (Cambridge, England), (32)(32), 3398-3400 (2007-11-21)
Chiral para-tert-butylcalix[4]arene bearing (S)-alpha-methylbenzylamine groups at lower rim only self-assembles with one of two enantiomers of 2,3-dibenzoyltartaric acid into coiled nanofibers and the coiled nanofibers only stack with the nanofibers having the same handedness to construct bigger ribbon-like fibers bearing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service