Merck
  • Interleukin-23 is sufficient to induce rapid de novo gut tumorigenesis, independent of carcinogens, through activation of innate lymphoid cells.
Interleukin-23 is sufficient to induce rapid de novo gut tumorigenesis, independent of carcinogens, through activation of innate lymphoid cells.
Mucosal immunology (2013-11-28)
I H Chan, R Jain, M S Tessmer, D Gorman, R Mangadu, M Sathe, F Vives, C Moon, E Penaflor, S Turner, G Ayanoglu, C Chang, B Basham, J B Mumm, R H Pierce, J H Yearley, T K McClanahan, J H Phillips, D J Cua, E P Bowman, R A Kastelein, D LaFace
ABSTRACT
Chronic inflammation has been associated with increased risk for developing gastrointestinal cancer. Interleukin-23 (IL-23) receptor signaling has been correlated with inflammatory bowel disease pathogenesis, as well as promotion of tumor growth. However, little is known about the relative potential for IL-23-directed causality in gut tumorigenesis. We report that IL-23 transgene expression was sufficient to induce rapid (3-4 weeks) de novo development of intestinal adenomas with 100% incidence. Initiation of tumorigenesis was independent of exogenous carcinogens, Helicobacter colonization, or pre-existing tumor-suppressor gene mutations. Tumorigenesis was mediated by Thy1(+)IL-23R(+) innate lymphoid cells (ILC3), in part, through IL-17 responses as tumor development was inhibited in RAG(-/-) × IL-17(-/-) double knockout mice. Remarkably, IL-23 initiation of tumorigenesis by resident ILCs consistently occurred before recruitment of conspicuous inflammatory infiltrates. Our results reveal an explicit role for IL-23-mediated initiation of gut tumorigenesis and implicate a key role for IL-23R(+) ILC3 in the absence of overt cellular infiltrate recruitment.
Sigma-Aldrich
3-Methylbutanol, for molecular biology, BioReagent, ≥98.5%
Sigma-Aldrich
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Supelco
3-Methylbutanol, analytical standard
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
3-Methyl-1-butanol, ACS reagent, ≥98.5%
Supelco
3-Methyl-1-butanol, analytical standard
Sigma-Aldrich
3-Methyl-1-butanol, reagent grade, 98%
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%