Skip to Content
Merck
All Photos(1)

Key Documents

74011

Supelco

(±)-Diclofop

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(RS)-2-[4-(2,4-Dichlorophenoxy)phenoxy]propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12Cl2O4
CAS Number:
Molecular Weight:
327.16
Beilstein:
2338390
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

CC(Oc1ccc(Oc2ccc(Cl)cc2Cl)cc1)C(O)=O

InChI

1S/C15H12Cl2O4/c1-9(15(18)19)20-11-3-5-12(6-4-11)21-14-7-2-10(16)8-13(14)17/h2-9H,1H3,(H,18,19)

InChI key

OOLBCHYXZDXLDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. The CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
(+/–)-Diclofop is a chiral herbicide that belongs to the chemical class of chiral aryloxyphenoxypropionate compounds. The actual herbicidal active ingredient― carboxylic acid, is released after application through hydrolysis of the ester. It is absorbed mainly through leaves and inhibits the biosynthesis of fatty acids by suppressing the activity of acetyl CoA carboxylase (ACCase). It is used for the post-emergence control of wild oats, wild millets, and other annual grass weeds commonly occurring in wheat, barley, rye, red fescue, and broad-leaved crops.
It was included on 1st June 2011 in Annex I of Directive 91/414/EEC by the European Commission Directive 2011/45/EU. It is authorized for use under EC Regulation No 1107/2009, as per the Commission Implementing Regulation (EU) No 540/2011, however it is a candidate for substitution.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques.
The (+/–)-Diclofop CRM can also be used as following:
  • To evaluate the likely enantioselective oxidative stress produced in Microcystis aeruginosa by diclofop acid
  • For analyzing the phytotoxic effects of diclofop acid enantiomers on the plant Arabidopsis thaliana
  • To study the enantioselective toxicity of diclofop acid on the non-target rice Xiushui 63 seedlings
  • In the chiral separation of diclofop-acid using one- and two- dimensional HPLC methods
  • To determine the enantioselective toxicity and degradation of diclofop in three algal cultures

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiyun Cai et al.
Journal of agricultural and food chemistry, 56(6), 2139-2146 (2008-03-06)
Aryloxyphenoxypropanoates are a class of chiral herbicides. They have a pair of enantiomers, only the R(+) form of which is herbicidally active. Diclofop, the model compound of these herbicides, is commercialized as the racemate of the ester form, diclofop-methyl, consisting
Vanesa Guillén-Casla et al.
Journal of agricultural and food chemistry, 56(7), 2303-2309 (2008-03-15)
Simple one- and two-dimensional high-performance liquid chromatography (HPLC) methods for the simultaneous enantiomeric determination of alkyloxyphenoxypropionic acid herbicides is presented. Compounds studied were ( R, S)-2-[4-(2,4-dichlorophenoxy)phenoxy]propionic acid (diclofop-acid) and ( R, S)-2-[4-(2,4-dichlorophenoxy)]methyl propionate (diclofop-methyl). Mobile phases necessary to separate their
Jing Ye et al.
Bulletin of environmental contamination and toxicology, 83(1), 85-91 (2009-05-20)
In this study, the acute toxicity (72-h EC50 values) of chiral diclofop acid towards rice Xiushui 63 seedlings and its effects on the Hill reaction activities of chloroplasts were determined. Significant differences were observed between the two enantiomers in 72-h
Jing Ye et al.
Aquatic toxicology (Amsterdam, Netherlands), 146, 12-19 (2013-11-19)
Enantioselective oxidative stress and toxin release from Microcystis aeruginosa after exposure to the chiral herbicide diclofop acid were investigated. Racemic diclofop acid, R-diclofop acid and S-diclofop acid induced reactive oxygen species (ROS) generation, increased the concentration of malondialdehyde (MDA), enhanced
Qiong Zhang et al.
Environmental science & technology, 46(15), 8405-8412 (2012-07-10)
Diclofop-methyl (DM) is a widely used chiral herbicide, which rapidly hydrolyzes to its major metabolite diclofop acid (DC) after application. With a carbon chiral center, DC not only is an important ingredient of herbicidal activity, but also has a long

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service