Skip to Content
Merck
All Photos(2)

Key Documents

51991

Supelco

Cypermethrin

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19Cl2NO3
CAS Number:
Molecular Weight:
416.30
Beilstein:
8444118
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas (argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)C(\C=C(\Cl)Cl)C1C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2

InChI

1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3

InChI key

KAATUXNTWXVJKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Nervous system, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hassane Adakal et al.
Experimental & applied acarology, 59(4), 483-491 (2012-09-13)
Traditional systems account for 95 % of the livestock produced in Burkina Faso. Tick infestation hampers livestock productivity in this area. However, little information exists on tick-control practices used by livestock farmers. We interviewed 60 stockbreeders working in traditional farming systems
Richard M Oxborough et al.
PloS one, 8(2), e55781-e55781 (2013-02-15)
Pyrethroid resistant Anopheles gambiae malaria vectors are widespread throughout sub-Saharan Africa and continued efficacy of pyrethroid ITNs is under threat. Chlorfenapyr is a promising pyrrole insecticide with a unique mechanism of action conferring no cross-resistance to existing public health insecticides.
Yan Fang Li et al.
Biomedical and environmental sciences : BES, 26(3), 201-208 (2013-02-22)
To evaluate effects of cypermethrin on the testis histology and testosterone, LH and FSH in adult male Sprague-Dawley rats. The intact adult male rats were randomly divided into five groups and were treated with cypermethrin at doses of 0, 7.5
Riin Muljar et al.
Journal of insect physiology, 58(11), 1469-1476 (2012-09-11)
Sublethal effects of pesticides in insects can be observed through physiological changes, which are commonly estimated by metabolic rate and respiratory patterns, more precisely by the patterns of discontinuous gas-exchange (DGE) cycles. The aim of the present research was to
M L Groner et al.
Journal of fish diseases, 36(3), 195-208 (2012-11-06)
Sea lice, Lepeophtheirus salmonis, are ectoparasites of farmed and wild salmonids. Infestations can result in significant morbidity and mortality of hosts in addition to being costly to control. Integrated pest management programmes have been developed to manage infestations, and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service