Skip to Content
Merck
All Photos(2)

Key Documents

71745

Supelco

Sodium ionophore IV

Selectophore, function tested

Synonym(s):

2,3:11,12-Didecalino-16-crown-5, 2,6,13,16,19-Pentaoxapentacyclo[18.4.4.47,12.01,20.07,12]dotriacontane, DD-16-C-5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O5
CAS Number:
Molecular Weight:
450.65
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

100
200

product line

Selectophore

form

powder

quality

function tested

storage temp.

2-8°C

SMILES string

C1COC23CCCCC2(CCCC3)OCCOCCOC45CCCCC4(CCCC5)OC1

InChI

1S/C27H46O5/c1-5-14-26-15-6-2-11-24(26,10-1)29-18-9-19-30-25-12-3-7-16-27(25,17-8-4-13-25)32-23-21-28-20-22-31-26/h1-23H2/t24-,25-,26+,27+

InChI key

ISSCXMQDZJDLSO-PVCJUVKBSA-N

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Sodium ionophore which shows an extremely high selectivity for Na over K (1000:1)

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Suzuki et al.
Analytical chemistry, 68(1), 208-215 (1996-01-01)
To develop an ionophore that is highly selective for sodium for use in an ion-selective electrode, we propose a model based on 16-crown-5 which has a cavity just the size of Na(+ )and has a "block" subunit to prevent complex
Vedrana Mikušević et al.
The Journal of general physiology, 151(12), 1357-1368 (2019-10-19)
KtrAB is a key player in bacterial K+ uptake required for K+ homeostasis and osmoadaptation. The system is unique in structure and function. It consists of the K+-translocating channel subunit KtrB, which forms a dimer in the membrane, and the
Judith A Heiny et al.
The Journal of general physiology, 151(9), 1146-1155 (2019-07-20)
Ion movements across biological membranes, driven by electrochemical gradients or active transport mechanisms, control essential cell functions. Membrane ion movements can manifest as electrogenic currents or electroneutral fluxes, and either process can alter the extracellular and/or intracellular concentration of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service