Skip to Content
Merck
All Photos(1)

Key Documents

71733

Supelco

Sodium ionophore II

Selectophore, function tested

Synonym(s):

ETH 157, N,N′-Dibenzyl-N,N′-diphenyl-1,2-phenylenedioxydiacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H32N2O4
CAS Number:
Molecular Weight:
556.65
Beilstein:
2930077
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

powder

quality

function tested

SMILES string

O=C(COc1ccccc1OCC(=O)N(Cc2ccccc2)c3ccccc3)N(Cc4ccccc4)c5ccccc5

InChI

1S/C36H32N2O4/c39-35(37(31-19-9-3-10-20-31)25-29-15-5-1-6-16-29)27-41-33-23-13-14-24-34(33)42-28-36(40)38(32-21-11-4-12-22-32)26-30-17-7-2-8-18-30/h1-24H,25-28H2

InChI key

PVWNDKPREJPMNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium ionophore II is commonly used sodium selective neutral carrier having 2.5 times more sensitivity towards sodium ions than to potassium ions. It can be used in ion-selective electrode preparation.
Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Neutral ionophore for liquid-membrane electrodes of high selectivity for Na+.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Na+ and K+ concentrations, extra-and intracellular voltages, and the effect of TTX in hypoxic rat hippocampal slices.
Muller, Michael, and George G. Somjen.
Journal of Neurophysiology, 83.2, 735-745 (2000)
W. Simon et al.
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 44, 613-613 (1975)
One-component room temperature vulcanizing-type silicone rubber-based sodium-selective membrane electrodes.
Poplawski, Michael E., et al.
Analytica Chimica Acta, 355.2, 249-257 (1997)
F. Deyhimi et al.
Fluid Phase Equilibria, 254, 18-27 (2007)
D. Ammann et al.
Ion and Enzyme Electrodes in Biology and Medicine (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service