Skip to Content
Merck
All Photos(2)

Documents

69686

Supelco

Shikimic acid

analytical standard

Synonym(s):

(3R,4S,5R)-(−)-3,4,5-Trihydroxy-1-cyclohexenecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O5
CAS Number:
Molecular Weight:
174.15
Beilstein:
2210055
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

98.0-102.0% (T)

optical activity

[α]/D -180.0±5.0°, c = 4 in H2O

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

185-187 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1CC(=C[C@@H](O)[C@H]1O)C(O)=O

InChI

1S/C7H10O5/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1,4-6,8-10H,2H2,(H,11,12)/t4-,5-,6-/m1/s1

InChI key

JXOHGGNKMLTUBP-HSUXUTPPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Shikimic acid has been used as a reference standard in the separation and determination of the analyte in fronds of brown seaweed, Cystoseira tamariscifolia using ultra high-performance liquid chromatography with ultraviolet detection (UHPLC-UV).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: ginkgo

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ecophysiological and metabolic responses to interactive exposure to nutrients and copper excess in the brown macroalga Cystoseira tamariscifolia.
Celis-Pla MSP, et al.
Marine Pollution Bulletin, 128, 214-222 (2018)
Kai Chen et al.
Bioresource technology, 119, 141-147 (2012-06-26)
Shikimic acid (SA) is an important metabolic intermediate with diverse commercial applications. In this work, antisense RNA interference and gene deletion were carried out to inactivate the aroK gene in an SA-producing Escherichia coli strain, DHPYA-T7. In this strain, the
Varun Rawat et al.
Organic & biomolecular chemistry, 10(20), 3988-3990 (2012-04-24)
A new enantioselective synthesis of the anti-influenza agent (-)-oseltamivir free base (7.1% overall yield; 98% ee) and (-)-methyl 3-epi-shikimate (16% overall yield; 98% ee) has been described from readily available raw materials. Sharpless asymmetric epoxidation and diastereoselective Barbier allylation of
Chloé Lemaître et al.
Journal of bacteriology, 196(7), 1343-1349 (2014-01-21)
The ability to capture iron is a challenge for most bacteria. The neonatal meningitis Escherichia coli strain S88 possesses several iron uptake systems, notably including siderophores. Transcriptional analysis of the ColV plasmid pS88 has shown strong induction of a previously
Amalia M Estévez et al.
Mini reviews in medicinal chemistry, 12(14), 1443-1454 (2012-07-26)
Shikimic acid, a natural compound is a key intermediate in the biosynthesis of amino acids. Consequently, this derivative is widely present in many plants and has interesting biological properties. But besides the pharmacological relevance of shikimic acid itself, it is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service