Skip to Content
Merck
All Photos(2)

Key Documents

260428

Sigma-Aldrich

3-Ethoxy-1,2-propanediol

98%

Synonym(s):

3-Ethoxypropane-1,2-diol, Glycerol 1-ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
120.15
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

222 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

SMILES string

CCOCC(O)CO

InChI

1S/C5H12O3/c1-2-8-4-5(7)3-6/h5-7H,2-4H2,1H3

InChI key

LOSWWGJGSSQDKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Ethoxy-1,2-propanediol (also known as glycerol α-ethyl ether) is primarily used as a solvent in various industries, including pharmaceuticals, cosmetics, and coatings. It can also be used as a substrate in the synthesis of cyclic phosphates for use in flame retardants.

Application

3-Ethoxy-1,2-propanediol (EGE) can be used as a crosslinking agent in the development of poly(glycidol) hydrogels for use in various biomedical applications such as tissue engineering and drug delivery. It is also used as a potential component in a bio-based solvent system for dissolving or processing nitrocellulose.

3-Ethoxy-1,2-propanediol can be used as:

  • An analyte protector in gas chromatographic analysis of pesticides.
  • An acceptor substrate in glycosylation reaction by sucrose phosphorylase.
It can also be used as a precursor in the Mitsunobu reaction of N-hydroxy phthalimide with poly(glycidol) and synthesis of 2-(3-ethoxy-2-hydroxypropoxy) isoindoline-1,3-dione.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven J Lehotay et al.
Journal of agricultural and food chemistry, 68(5), 1468-1479 (2020-01-17)
In this study, sample processing of bulk commodities using an efficient one-step comminution procedure with liquid nitrogen (LN2) was devised and assessed in the analysis of pesticide residues in fruits and vegetables. LN2 was added to the fresh samples from
Lucia Ivorra et al.
Chemosphere, 235, 662-669 (2019-07-06)
Uptake and depuration kinetics of 4,4'-dichlorobenzophenone (main metabolite of dicofol) in the edible clam Meretrix meretrix were evaluated through a mesocosm experiment. M. meretrix was exposed to different dicofol concentrations (environmental concentration, D1 = 50 ng/L; supra-environmental concentration, D2 = 500 ng/L) for 15 days, followed by
Jian Du et al.
Chemosphere, 167, 247-254 (2016-10-12)
Persistent organic pollutants in human milk (HM) at high levels are considered to be detrimental to the breastfed infant. To determine the pesticide concentration in HM, a pilot cross-sectional study of 40 Western Australian (WA) women was carried out. Gas
Jian Du et al.
Scientific reports, 6, 38355-38355 (2016-12-08)
The presence of pesticides in human milk (HM) is of great concern due to the potential health effects for the breastfed infant. To determine the relationships between HM pesticides and infant growth and development, a longitudinal study was conducted. HM
Lijun Han et al.
Journal of agricultural and food chemistry, 66(20), 4986-4996 (2017-11-15)
In this study, analytical results were compared when using different approaches to bulk food sample comminution, consisting of a vertical chopper (Blixer) at room temperature and dry ice cryogenic conditions, followed by further subsample processing (20 g) using liquid nitrogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service