Skip to Content
Merck
All Photos(2)

Documents

68824

Supelco

Cetyl alcohol

analytical standard

Synonym(s):

1-Hexadecanol, Palmityl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15OH
CAS Number:
Molecular Weight:
242.44
Beilstein:
1748475
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

190 °C/15 mmHg (lit.)

mp

49-51 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCCCCCO

InChI

1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

BXWNKGSJHAJOGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cetyl alcohol is a long chain alcohol, which can find applications as an emulsifying agent, emollient, viscosity increasing agent, foam booster and opacifying agent in pharmaceuticals and cosmetics.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Cetyl alcohol can undergo reaction with arachidic acid to form cetyl arachidate.
Cetyl alcohol may be used as an excipient for the determination of acyclovir in pharmaceutical preparations and tranexamic acid in cosmetic products by chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Holmberg K
Novel Surfactants: Preparation Applications And Biodegradability, Second Edition, Revised And Expanded (Vol 114 of Surfactant Science), (2) (2003)
Winter R
A Consumer's Dictionary of Cosmetic Ingredients: Complete Information About the Harmful and Desirable Ingredients in Cosmetics and Cosmeceuticals (2010)
Smolinske.CS
CRC Handbook of Food, Drug, and Cosmetic Excipients (1992)
Determination of tranexamic acid in cosmetic products by high-performance liquid chromatography coupled with barrel plating nickel electrode
Shih Y, et al.
Journal of Pharmaceutical and Biomedical Analysis, 48(5), 1446-1450 (2008)
TLC determination of acyclovir in pharmaceutical preparations, and validation of the method used
Sia T, et al.
Journal of Planar Chromatography--Modern TLC, 15(1), 42-45 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service