Skip to Content
Merck

Skip To

60740

Silica gel Inorganic Sorbent

high-purity grade (puriss), 70-230 mesh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
SB.52
EC Number:
231-545-4
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Silica gel, high-purity grade (puriss), pore size 60 Å, 70-230 mesh, for column chromatography

grade

high-purity grade (puriss)

Quality Segment

description

activity acc. to Brockmann and Schodder 2 - 3

form

powder

manufacturer/tradename

Sigma-Aldrich

technique(s)

LPLC: suitable

surface area

500 m2/g

matrix

Silica

matrix active group

silica

particle size

63-200 μm, 70-230 mesh

pore size

0.8 cm3/g pore volume, 60 Å

pH

6.0-7.5 (25 °C, 10% in H2O)

bp

2230 °C

mp

>1600 °C

anion traces

chloride (Cl-): ≤100 mg/kg, nitrate (NO3-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Cu: ≤5 mg/kg, Fe: ≤20 mg/kg, Pb: ≤5 mg/kg, Zn: ≤5 mg/kg

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

General description

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium (III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction

Application

Silica gel may be used in separation of single-walled carbon nanotube (SWNT) by simple filtration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
607456074660736
description

high-purity grade (puriss), pore size 60 Å, 70-230 mesh, for column chromatography

description

high-purity grade, 90Å, 70-230 mesh, suitable for column chromatography

description

high purity, 90Å, 35-70 mesh, suitable for column chromatography

description

high-purity grade, 40, 35-70 mesh, suitable for column chromatography

separation technique

hydrophilic interaction (HILIC)

separation technique

hydrophilic interaction (HILIC)

separation technique

hydrophilic interaction (HILIC)

separation technique

hydrophilic interaction (HILIC)

particle size

70-230 mesh, 63-200 μm

particle size

0.063-0.200 mm (70-230 mesh ASTM), 70-230 mesh

particle size

35-70 mesh, 0.2-0.5 mm (35-70 mesh ASTM)

particle size

35-70 mesh, 0.2-0.5 mm (35-70 mesh ASTM)

pore size

60 Å, 0.8 cm3/g pore volume

pore size

90 Å mean pore size

pore size

90 Å mean pore size

pore size

90 Å mean pore size

matrix

Silica

matrix

Silica

matrix

Silica

matrix

Silica

matrix active group

silica

matrix active group

silica

matrix active group

silica

matrix active group

silica

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable


Still not finding the right product?

Explore all of our products under Silica gel


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters