Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
SB.52
EC Number:
231-545-4
MDL number:
Product Name
Silica gel, high-purity grade, 90Å, 70-230 mesh, suitable for column chromatography
grade
high-purity grade
Quality Segment
form
powder or crystals
technique(s)
LPLC: suitable
surface area
500 m2/g , approx.
matrix
Silica
matrix active group
silica
particle size
0.063-0.200 mm (70-230 mesh ASTM), 70-230 mesh
pore size
90 Å mean pore size
bp
2230 °C
mp
>1600 °C
suitability
suitable for column chromatography
separation technique
hydrophilic interaction (HILIC)
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
General description
Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium (III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction
Still not finding the right product?
Explore all of our products under Silica gel
Storage Class
11 - Combustible Solids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.