Skip to Content
Merck
All Photos(1)

Key Documents

523356

Supelco

Poly(2-vinylpyridine)

analytical standard, average Mw 159,000 (Typical), average Mn 152,000 (Typical)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

mol wt

average Mn 152,000 (Typical)
average Mw 159,000 (Typical)

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

InChI

1S/C7H7N/c1-2-7-5-3-4-6-8-7/h2-6H,1H2

InChI key

KGIGUEBEKRSTEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleksandr Trotsenko et al.
Langmuir : the ACS journal of surfaces and colloids, 28(14), 6037-6044 (2012-03-15)
Conformations of cationic polyelectrolytes (PEs), a weak poly(2-vinylpyridine) (P2VP) and a strong poly(N-methyl-2-vinylpyridinium iodide) (qP2VP), adsorbed on mica from saline solutions in the presence of counterions of different valences are studied using in situ atomic force microscopy (AFM). Quantitative characteristics
Joseph P Cook et al.
Advances in colloid and interface science, 147-148, 67-73 (2008-12-27)
The effect of perchlorate ions as counterions in the acid-induced swelling of poly(2-vinylpyridine) microgel particles was investigated. The pH was modified with perchloric, hydrochloric and hydrobromic acids and the particle diameter measured using dynamic light scattering and the viscosity monitored
Sina Burkert et al.
Langmuir : the ACS journal of surfaces and colloids, 26(3), 1786-1795 (2009-09-22)
Protein adsorption, as the primary process occurring when a foreign surface comes into contact with a biosystem, was studied on thin polymer brush films consisting of poly(N-isopropylacrylamide) (PNIPAAm) and poly(2-vinylpyridine) (P2VP). These films were prepared by the "grafting to" method.
Astrid Drechsler et al.
Langmuir : the ACS journal of surfaces and colloids, 28(44), 15555-15565 (2012-10-13)
AFM force measurements have been performed to study the influence of the pH value and salt concentration on the interactions between poly(2-vinyl pyridine) brushes and microsized silica spheres, focusing on attractive and adhesion forces. It was found that the interaction
Lung-Chi Chen et al.
Chemical communications (Cambridge, England), 48(30), 3668-3670 (2012-03-08)
Stereoregular vinyl polymers, poly(2-vinyl pyridine)s (P2VPs), were synthesized to examine the tacticity effect on the induced circular dichroism (ICD) via association with chiral acids. The ICD was found to be strongly dependent on the isotacticity of the P2VPs and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service