Skip to Content
Merck
All Photos(2)

Key Documents

472352

Sigma-Aldrich

Poly(4-vinylpyridine)

average Mw ~160,000

Synonym(s):

4-Vinylpyridine polymer, Poly(p -vinylpyridine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C7H7N)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mw ~160,000

Quality Level

transition temp

Tg 142 °C (onset, annealed)

solubility

DMF, acetic acid and lower alcohols: soluble

SMILES string

n1ccc(cc1)C=C

InChI

1S/C7H7N/c1-2-7-3-5-8-6-4-7/h2-6H,1H2

InChI key

KFDVPJUYSDEJTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(4-vinylpyridine) can be used:
  • To prepare metal-containing polymer, applicable as nonlinear optically active material.
  • For the removal of perchlorate from an aqueous solution.
It can also be incorporated into active layers of organic solar cells to enhance power conversion efficiency.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Rita Rodrigues et al.
Materials (Basel, Switzerland), 7(12), 8189-8196 (2014-12-22)
We report on the efficiency improvement of organic solar cells (OPVs) based on the low energy gap polyfluorene derivative, APFO-3, and the soluble C60 fullerene PCBM, upon addition of a residual amount of poly (4-vinylpyridine) (PVP). We find that the
Zenghe Liu et al.
Analytical chemistry, 84(7), 3403-3409 (2012-03-20)
Continuous glucose measurement has improved the treatment of type 1 diabetes and is typically provided by externally powered transcutaneous amperometric sensors. Self-powered glucose sensors (SPGSs) could provide an improvement over these conventionally powered devices, especially for fully implanted long-term applications
Lin-feng Zhou et al.
Journal of chromatography. A, 1256, 15-21 (2012-08-14)
A simple and facile needle-adapter was designed for constructing manual on-line polymer monolith microextraction-high performance liquid chromatography (PMME-HPLC). A capillary poly(4-vinylpyridine-co-ethylene glycol dimethacrylate) [poly(VP-co-EGDMA)] monolith was prepared by in situ polymerization, using 4-vinylpyridine (VP) and ethylene glycol dimethacrylate (EGDMA) as
Biomimetic surface coatings for atmospheric water capture prepared by dewetting of polymer films.
Stuart C Thickett et al.
Advanced materials (Deerfield Beach, Fla.), 23(32), 3718-3722 (2011-07-19)
Sampa Saha et al.
ACS applied materials & interfaces, 3(8), 3042-3048 (2011-07-07)
Atom transfer radical polymerization (ATRP) is commonly used to grow polymer brushes from Au surfaces, but the resulting film thicknesses are usually significantly less than with ATRP from SiO(2) substrates. On Au, growth of poly(methyl methacrylate) (PMMA) blocks from poly(tert-butyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service