Skip to Content
Merck
All Photos(2)

Key Documents

50787

Supelco

Bisoprolol

analytical standard

Synonym(s):

1-{4-{[2-(1-Methylethoxy)ethoxy]methyl}phenoxy}-3-[(1-methylethyl)amino]-2-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H31NO4
CAS Number:
Molecular Weight:
325.44
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)OCCOCC1=CC=C(OCC(O)CNC(C)C)C=C1

InChI

1S/C18H31NO4/c1-14(2)19-11-17(20)13-23-18-7-5-16(6-8-18)12-21-9-10-22-15(3)4/h5-8,14-15,17,19-20H,9-13H2,1-4H3

InChI key

VHYCDWMUTMEGQY-UHFFFAOYSA-N

Gene Information

human ... ADRB1(153)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - STOT RE 2

Target Organs

Heart

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An S De Vriese et al.
Journal of the American Society of Nephrology : JASN, 31(1), 186-196 (2019-11-11)
Vitamin K antagonists (VKAs), although commonly used to reduce thromboembolic risk in atrial fibrillation, have been incriminated as probable cause of accelerated vascular calcification (VC) in patients on hemodialysis. Functional vitamin K deficiency may further contribute to their susceptibility for
Dong Hyun Yang et al.
Radiology, 272(3), 665-673 (2014-04-05)
To determine whether C-reactive protein is associated with the type of coronary plaque seen at computed tomographic (CT) angiography. The institutional review board approved this retrospective study, and the need for informed consent was waived. C-reactive protein levels were measured
Augustine N Odili et al.
Trials, 13, 59-59 (2012-05-19)
The epidemic surge in hypertension in sub-Saharan Africa is not matched by clinical trials of antihypertensive agents in Black patients recruited in this area of the world. We mounted the Newer versus Older Antihypertensive agents in African Hypertensive patients (NOAAH)
Gyung-Min Park et al.
Lipids in health and disease, 19(1), 7-7 (2020-01-16)
Atherosclerotic cardiovascular (CV) events commonly occur in individuals with a low CV risk burden. This study evaluated the ability of the triglyceride glucose (TyG) index to predict subclinical coronary artery disease (CAD) in asymptomatic subjects without traditional CV risk factors
Gyung-Min Park et al.
Scientific reports, 9(1), 15306-15306 (2019-10-28)
The benefit of a high level of high-density lipoprotein cholesterol (HDL-C) against coronary atherosclerosis risk after achieving optimal glycemic control (OGC) in diabetics remains uncertain. We aimed to evaluate the association between HDL-C and obstructive coronary artery disease (CAD) according

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service