Skip to Content
Merck
All Photos(2)

Documents

49761

Supelco

Spermidine

analytical standard

Synonym(s):

1,8-Diamino-4-azaoctane, N-(3-Aminopropyl)-1,4-diaminobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3NH(CH2)4NH2
CAS Number:
Molecular Weight:
145.25
Beilstein:
1698591
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (NT)
≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.479 (lit.)

density

0.925 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

NCCCCNCCCN

InChI

1S/C7H19N3/c8-4-1-2-6-10-7-3-5-9/h10H,1-9H2

InChI key

ATHGHQPFGPMSJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Spermidine is a naturally occurring polyamine compound commonly found in food. It is essential for cell growth and survival.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Spermidine may be used as one of the reference standard for the determination of spermidine in biological samples by high performance liquid chromatography-electrospray ionization–ion trap tandem mass spectrometry (HPLC–ESI–ITMS/MS).

Biochem/physiol Actions

Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA; may be used for purification of DNA binding proteins. Stimulates T4 polynucleotide kinase activity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Caution

Solutions should be sterile-filtered, not autoclaved, if sterile solution is necessary. Spermidine deaminates with time; solutions should be stored frozen. Prepare new solutions frequently.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micro-scale strategy to detect spermine and spermidine by MALDI?TOF MS in foods and identification of apoptosis-related proteins by nano-flow UPLC?MS/MS after treatment with spermine and spermidine
Su H H, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 978, 131-137 (2015)
Determination of putrescine, cadaverine, spermidine and spermine in different chemical matrices by high performance liquid chromatography?electrospray ionization?ion trap tandem mass spectrometry (HPLC?ESI?ITMS/MS).
Ibarra A A G, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1002, 176-184 (2015)
Anita Nielsen et al.
Marine drugs, 10(11), 2584-2595 (2012-12-04)
Staphylococcus aureus is a serious human pathogen that employs a number of virulence factors as part of its pathogenesis. The purpose of the present study was to explore marine bacteria as a source of compounds that modulate virulence gene expression
Maria Kostakioti et al.
Cold Spring Harbor perspectives in medicine, 3(4), a010306-a010306 (2013-04-03)
Biofilm formation constitutes an alternative lifestyle in which microorganisms adopt a multicellular behavior that facilitates and/or prolongs survival in diverse environmental niches. Biofilms form on biotic and abiotic surfaces both in the environment and in the healthcare setting. In hospital
Eugenia Morselli et al.
Aging, 1(12), 961-970 (2010-02-17)
Although autophagy has widely been conceived as a self-destructive mechanism that causes cell death, accumulating evidence suggests that autophagy usually mediates cytoprotection, thereby avoiding the apoptotic or necrotic demise of stressed cells. Recent evidence produced by our groups demonstrates that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service