Skip to Content
Merck

41346

2-Phenethylamine

analytical standard

Synonym(s):

β-Phenylethylamine, 2-Phenylethylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5CH2CH2NH2
CAS Number:
Molecular Weight:
121.18
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
200-574-4
Beilstein/REAXYS Number:
507488
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

4.18 (vs air)

assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.533 (lit.), n20/D 1.534

bp

197-200 °C (lit.)

density

0.962 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

NCCc1ccccc1

InChI

1S/C8H11N/c9-7-6-8-4-2-1-3-5-8/h1-5H,6-7,9H2

InChI key

BHHGXPLMPWCGHP-UHFFFAOYSA-N

General description

2-Phenethylamine belongs to the class of biogenic amines, commonly found in a variety of wines, food products and also in dietary supplements.
2-Phenethylamine is commonly known as a substructure of many biological active compounds. It can also be used in the synthesis of 1,2,3,4-tetrahydroisoquinoline derivatives, via Pictet-Spengler reaction, which is one of the key reactions for construction of the isoquinoline skeleton, involving a naturally occurring bioactive compound.

Application

2-Phenethylamine may be used as an analytical reference standard for the quantification of the analyte in wine samples and food products using high-performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 1

flash_point_f

177.8 °F - closed cup

flash_point_c

81 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Synthesis of Photoreactive 2?Phenethylamine Derivatives?Synthesis of Adenosine Derivatives Enabling Functional Analysis of Adenosine Receptors by Photoaffinity Labeling
Murai Y, et al.
European Journal of Organic Chemistry, 2013, 2428-2433 (2013)
Investigation of biogenic amines in sufu (furu): A Chinese traditional fermented soybean food product
Guan F-R, et al.
Food Control, 31(2), 345-352 (2013)
Stereoselectivity of Superacid-Catalyzed Pictet? Spengler Cyclization Reactions
Nakamura S, et al.
Organic Letters, 5, 2087-2090 (2003)



Global Trade Item Number

SKUGTIN
41346-1ML-F04061832081748
41346-5ML-F04061832081793