Skip to Content
Merck
All Photos(1)

Documents

46473

Supelco

Morantel tartrate hydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N2O6S · xH2O
CAS Number:
Molecular Weight:
370.42 (anhydrous basis)
Beilstein:
8739506
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCCN=C1\C=C\c2sccc2C

InChI

1S/C12H16N2S.C4H6O6.H2O/c1-10-6-9-15-11(10)4-5-12-13-7-3-8-14(12)2;5-1(3(7)8)2(6)4(9)10;/h4-6,9H,3,7-8H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10);1H2/b5-4+;;/t;1-,2-;/m.1./s1

InChI key

XMDWLUCAURURDF-CIIIEQSVSA-N

General description

Chemical structure: tetrahydropyrimidine

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Alonso-Villalobos et al.
Journal of veterinary medicine. B, Infectious diseases and veterinary public health, 47(1), 1-8 (2000-04-26)
The secretion of acetylcholinesterase (AChE) by female and male Heligmosomoides polygyrus was studied in different in vitro culture media. AChE secretion was increased in the presence of fetal calf serum or bovine serum albumin (BSA). In the absence of crowding
Christopher T Konek et al.
Journal of the American Chemical Society, 127(45), 15771-15777 (2005-11-10)
It is now known that the untreated discharge of pharmaceuticals into the environment can impact human health and development and lead to increased drug resistance in biota. Here, we present the first direct interface-specific studies that address the mobility of
M M Martínez-Grueiro
Parasitology research, 88(10), 946-949 (2002-09-05)
Acid phosphatase (AP) activity was detected in 24 h culture media from adult Heligmosomoides polygyrus. Female and male excretion/secretion products showed similar specific activity. For both, the AP had a pH optimum of 4.0 and was inhibited by sodium fluoride
V Raymond et al.
Neuroscience, 101(3), 785-791 (2000-12-13)
Two homomer-forming nicotinic acetylcholine receptor subunits with 47% identity in their amino acid sequences were employed to compare the actions of cholinergic anthelmintics and ivermectin on expressed vertebrate and nematode nicotinic receptors of known molecular composition. Voltage-clamp electrophysiology was used
John Jensen et al.
Environmental toxicology and chemistry, 28(2), 316-323 (2008-09-25)
Soil invertebrates in arable land are potentially exposed to veterinary medicines excreted by husbandry. The toxicity of three widely used pharmaceuticals was therefore investigated with the use of common soil invertebrates exposed in the laboratory in single- or two-species test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service