Skip to Content
Merck
All Photos(1)

Key Documents

SML0123

Sigma-Aldrich

Deflazacort

≥98% (HPLC)

Synonym(s):

(11β,16β)-21-(acetyloxy)-11-hydroxy-2′-methyl-5′H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 11β,21-dihydroxy-2′-methyl-5′βH-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione 2′-acetate, DL-458-IT, L-5458, azacort, oxazacort, pregna-1,4-diene-11β,21-diol-3,20-dione[17α,16α-d]-2′-methyloxazoline 21-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H31NO6
CAS Number:
Molecular Weight:
441.52
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)OCC(=O)[C@@]12N=C(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]23C

InChI

1S/C25H31NO6/c1-13-26-25(20(30)12-31-14(2)27)21(32-13)10-18-17-6-5-15-9-16(28)7-8-23(15,3)22(17)19(29)11-24(18,25)4/h7-9,17-19,21-22,29H,5-6,10-12H2,1-4H3/t17-,18-,19-,21+,22+,23-,24-,25+/m0/s1

InChI key

FBHSPRKOSMHSIF-GRMWVWQJSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Deflazacort is an anti-inflammatory and immunosuppressant glucocorticoid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Mayo et al.
Neuromuscular disorders : NMD, 22(12), 1040-1045 (2012-07-25)
Quality of life in Duchenne Muscular Dystrophy (DMD) has improved significantly with corticosteroid treatment. However, corticosteroids decrease bone mass and increase vertebral fragility fracture risk. We report on bone health in 39 boys with DMD on long-term deflazacort (0.9 mg/kg/day)
Eric P Hoffman et al.
Physical medicine and rehabilitation clinics of North America, 23(4), 821-828 (2012-11-10)
Although prednisone has never been formally approved for use in Duchenne muscular dystrophy (DMD) by regulatory agencies, its efficacy has been confirmed in trials dating from the 1980s. There is a strong need for optimization of both specific type of
Maggie C Walter et al.
Orphanet journal of rare diseases, 8, 26-26 (2013-02-15)
Dysferlinopathies are autosomal recessive disorders caused by mutations in the dysferlin (DYSF) gene encoding the dysferlin protein. DYSF mutations lead to a wide range of muscular phenotypes, with the most prominent being Miyoshi myopathy (MM) and limb girdle muscular dystrophy
Carlo Fusco et al.
European journal of paediatric neurology : EJPN : official journal of the European Paediatric Neurology Society, 16(4), 373-378 (2011-12-27)
To determine efficacy and safety of corticosteroid treatment in patients with severe Sydenham's chorea paralytic form. This is a 4 years observational study on ten patient with severe paralytic form of Sydenham's chorea unresponsive to neuroleptics and antiepileptics agents, treated
Corticosteroids can reduce the severity of scoliosis in Duchenne muscular dystrophy.
Anne M Connolly et al.
The Journal of bone and joint surgery. American volume, 95(12), e86-e86 (2013-06-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service