Skip to Content
Merck
All Photos(1)

Documents

45677

Supelco

Terbutryn

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19N5S
CAS Number:
Molecular Weight:
241.36
Beilstein:
611817
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(NC(C)(C)C)nc(SC)n1

InChI

1S/C10H19N5S/c1-6-11-7-12-8(15-10(2,3)4)14-9(13-7)16-5/h6H2,1-5H3,(H2,11,12,13,14,15)

InChI key

IROINLKCQGIITA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terbutryn is primarily used to control a wide variety of annual grass and broadleaf weed species. This preemergence and postemergence s-triazine herbicide inhibits root development by interrupting mitosis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Terbutryn may be used as a reference standard for the determination of terbutryn in wastewater and soil by micellar liquid chromatography.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of diuron, terbuthylazine, and terbutryn in wastewater and soil by micellar liquid chromatography.
Pitarch-Andres S, et al.
Analytical and Bioanalytical Chemistry, 409(8), 2037-2049 (2017)
In vitro testing for genotoxicity of the herbicide terbutryn: cytogenetic and primary DNA damage.
Moretti M, et al.
Toxicology in vitro, 16(1), 81-88 (2002)
Effect of Pendimethalin, Trifluralin and Terbutryn on Lolium multiflorum growing with barley during pre-emergence stage.
Alshallash KS.
Annals of Agricultural Science, 59(2), 239-242 (2014)
Carmen Rioboo et al.
Aquatic toxicology (Amsterdam, Netherlands), 94(3), 229-237 (2009-08-15)
Very little is known about growth and proliferation in relation to the cell cycle regulation of algae. The lack of knowledge is even greater when referring to the potential toxic effects of pollutants on microalgal cell division. To assess the
K Marja Talja et al.
Journal of agricultural and food chemistry, 56(24), 11962-11968 (2008-12-05)
The degradation of pesticides atrazine and terbutryn was investigated under aerobic and anaerobic conditions in the northern boreal region subsurface deposits and sterilized controls from the depths of 6.3-21.0 m below the surface and 1.2-16.9 m below the groundwater table.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service