Skip to Content
Merck
All Photos(1)

Key Documents

45678

Supelco

Terbuthylazine

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16ClN5
CAS Number:
Molecular Weight:
229.71
Beilstein:
3951137
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

solid phase extraction (SPE): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(Cl)nc(NC(C)(C)C)n1

InChI

1S/C9H16ClN5/c1-5-11-7-12-6(10)13-8(14-7)15-9(2,3)4/h5H2,1-4H3,(H2,11,12,13,14,15)

InChI key

FZXISNSWEXTPMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Application


  • Pesticide distribution and atmospheric persistence: An extensive study reveals the widespread distribution of pesticides, including Terbuthylazine, in the European atmosphere, challenging assumptions about their degradability. This highlights the need for enhanced regulatory measures and monitoring strategies to mitigate environmental and health risks associated with persistent atmospheric contaminants (Mayer et al., 2024).

  • Herbicide impact on non-target plant species: Research evaluates the early responses of Solanum nigrum L. to herbicide treatments involving Terbuthylazine, investigating changes in antioxidant activity, osmolyte concentrations, and chlorophyll fluorescence. This study contributes to understanding the selective action of Terbuthylazine and its broader ecological impacts, providing insights for developing more targeted and sustainable herbicidal applications (Hassannejad et al., 2023).

  • Human health risk from groundwater herbicide contamination: A human health risk assessment approach is applied to study the occurrence of herbicides, including Terbuthylazine, in the groundwater of Northern Greece. The findings emphasize the importance of continuous environmental monitoring and the development of mitigation strategies to ensure water safety and public health (Parlakidis et al., 2022).

  • Seasonal distribution of pesticide residues in water bodies: A study on the seasonal distribution of multiclass pesticide residues in surface waters of northwest Croatia includes Terbuthylazine, discussing the implications for aquatic ecosystems and the effectiveness of current water treatment practices. This research is crucial for assessing the environmental persistence and ecological risks of such chemicals (Fingler et al., 2021).

  • Direct pesticide exposure in conservation areas: An investigation into direct pesticide exposure, including Terbuthylazine, in nature conservation areas in Germany, underscores the challenges of protecting vulnerable ecosystems from agricultural chemical intrusion. The study highlights the critical need for stringent environmental protections and pesticide use regulations to preserve biodiversity (Brühl et al., 2021).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Veronika Bókony et al.
Environmental pollution (Barking, Essex : 1987), 260, 114078-114078 (2020-02-12)
Despite intensive ecotoxicological research, we still know relatively little about the ecological impacts of many environmental contaminants. Filling these knowledge gaps is particularly important regarding amphibians, because they play significant roles in freshwater and terrestrial ecosystems, and their populations are
Rosa Mercadante et al.
Toxicology letters, 210(2), 169-173 (2011-12-08)
Terbuthylazine (TBA) is an herbicide widely used in corn cultivation. Herein we evaluate the measurement of hair TBA as biomarkers of exposure. Five Sprague Dawley rats were gavaged with TBA for 3 days, and then the back hair was shaved
Rosa Mercadante et al.
Analytical and bioanalytical chemistry, 404(3), 875-886 (2012-07-04)
Terbuthylazine (TBA) is a widely applied herbicide and an environmental contaminant. Following its use, humans, such as agricultural workers and rural residents, may be exposed. An isotope-dilution liquid chromatography coupled to electrospray-tandem mass spectrometry method for the determination of TBA
Alex Laini et al.
The Science of the total environment, 438, 312-318 (2012-09-29)
Herbicides reduce the diversity of flora and fauna in freshwater ecosystems and also contaminate groundwater due to leaching. Herbicide contamination can be a serious threat for all groundwater-dependent ecosystems (GDE), altering their chemical and biological quality. Successful management to protect
Dubravka Spoljaric et al.
Aquatic toxicology (Amsterdam, Netherlands), 105(3-4), 552-558 (2011-09-23)
Oxidative stress, i.e. excessive production of reactive oxygen species (ROS), leads to lipid peroxidation and to formation of reactive aldehydes (e.g. 4-hydroxy-2-nonenal; HNE), which act as second messengers of free radicals. It was previously shown that herbicides can induce ROS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service